Skip to Content
Merck
All Photos(1)

Key Documents

SRP3087

Sigma-Aldrich

IL-21 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

IL-21 Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.5 ng/mL ED50

mol wt

15.4 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL21(59067)

General description

IL21 (interleukin 21) is a member of the γ-chain type I cytokine family, and a four-α-helix bundle pleiotropic cytokine. This gene is localized to human chromosome 4q26–q27. Although, it is predominantly produced by T follicular helper (TFH) and T helper-17 (TH17) cells, it is also produced by CD4+ T cells, neutrophils and natural killer cells. Under certain conditions, it is also produced by CD8+ T cells. Recombinant human IL-21 is a 15.0 kDa protein consisting of 132 amino acid residues.

Biochem/physiol Actions

IL21 (interleukin 21) functions on its heterodimeric receptor IL21R, which is expressed widely on immune and non-immune cells. Therefore, IL21 exerts its effects on a wide range of cell types and has a pleiotropic role in immune response regulation. This protein activates Janus kinase (JAK)-signal transducer and activator of transcription (STAT) signaling pathway and functions through mitogen-activated protein kinase (MAPK) and phosphoinositide 3-kinase (PI3K)-AKT signaling pathways. Studies show that in rheumatoid arthritis fibroblast-like synoviocytes (RA-FLS), IL21 facilitates migration, invasion and production of MMPs (MMP-2, MMP-3, MMP-9, MMP-13) (matrix metalloproteinases). Therefore, it has potential as a therapeutic target in the treatment of RA. Studies show that CD3+TCRvβ11+ NK (natural killer) T cells isolated from a local Mycobacterium tuberculosis infection site produce IL21, which in turn assists B cells to produce and release IgG (immunoglobulin) and IgA, to help fight the infection.

Sequence

MQDRHMIRMR QLIDIVDQLK NYVNDLVPEF LPAPEDVETN CEWSAFSCFQ KAQLKSANTG NNERIINVSI KKLKRKPPST NAGRRQKHRL TCPSCDSYEK KPPKEFLERF KSLLQKMIHQ HLSSRTHGSE DS

Physical form

Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin-21 induces migration and invasion of fibroblast-like synoviocytes from patients with rheumatoid arthritis.
Xing R, et al.
Clinical and Experimental Immunology, 184(2), 147-158 (2016)
Biological effects of IL-21 on different immune cells and its role in autoimmune diseases.
Gharibi T, et al.
Immunobiology, 221(2), 357-367 (2016)
Antigen-specific human NKT cells from tuberculosis patients produce IL-21 to help B cells for the production of immunoglobulins.
Wu C, et al.
Oncotarget, 6(30), 28633-28645 (2015)
Julia Parrish-Novak et al.
Journal of leukocyte biology, 72(5), 856-863 (2002-11-14)
Interleukin (IL)-21 was recently discovered using a functional cloning approach based on expression of its receptor. It is similar in domain organization and primary sequence to IL-2 and IL-15. Like these cytokines, IL-21 uses the common gamma chain of the
Hamza Loucif et al.
Autophagy, 18(6), 1256-1273 (2021-10-07)
The maintenance of a strong IL21 production in memory CD4 T cells, especially in HIV-1-specific cells, represents a major correlate of natural immune protection against the virus. However, the molecular mechanisms underlying IL21 production during HIV-1 infection, which is only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service