Skip to Content
Merck
All Photos(1)

Key Documents

B2680

Sigma-Aldrich

BMP4, human

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture

Synonym(s):

BMP-4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

product name

Bone Morphogenetic Protein 4 human, >95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

10-30 ng/mL ED50

mol wt

monomer calculated mol wt 13 kDa
~22 kDa by SDS-PAGE

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

technique(s)

cell culture | mammalian: suitable

impurities

<1 EU/mg Endotoxin level (LAL test)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... BMP4(652)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cellular responses to BMP-4 are mediated by the formation of hetero-oligomeric complexes of type I and type II serine/threonine kinase receptors. BMPs stimulate angiogenesis by inducing the production of VEGF A by osteoblasts. Mature human and mouse BMP-4 demonstrates 98% and 100% homology, respectively, to mature rat BMP-4.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoroacetic acid containing 50 μg of bovine serum albumin per 1 μg of cytokine.

Analysis Note

The biological activity is measured by its ability to induce alkaline phosphatase production by C2C12 myoblasts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

M Kawabata et al.
Cytokine & growth factor reviews, 9(1), 49-61 (1998-08-28)
Bone morphogenetic proteins (BMPs) are multifunctional cytokines, which are members of the transforming growth factor-beta (TGF-beta) superfamily. Activities of BMPs are extracellularly regulated by BMP-binding proteins, Noggin and Chordin. BMPs bind to two different types of serine-threonine kinase receptors, type
Martine M L Deckers et al.
Endocrinology, 143(4), 1545-1553 (2002-03-19)
During bone formation and fracture healing there is a cross-talk between endothelial cells and osteoblasts. We previously showed that vascular endothelial growth factor A (VEGF-A) might be an important factor in this cross-talk, as osteoblast-like cells produce this angiogenic factor
Thorsten Pfirrmann et al.
Human molecular genetics, 24(11), 3119-3132 (2015-02-26)
Chordin-Like 1 (CHRDL1) mutations cause non-syndromic X-linked megalocornea (XMC) characterized by enlarged anterior eye segments. Mosaic corneal degeneration, presenile cataract and secondary glaucoma are associated with XMC. Beside that CHRDL1 encodes Ventroptin, a secreted bone morphogenetic protein (BMP) antagonist, the
Meike Simann et al.
Bone, 78, 102-113 (2015-05-12)
Heparins are broadly used for the prevention and treatment of thrombosis and embolism. Yet, osteoporosis is considered to be a severe side effect in up to one third of all patients on long-term treatment. However, the mechanisms underlying this clinical
Ling Qin et al.
Life sciences, 131, 37-43 (2015-04-22)
Hyperphosphatemia is an independent risk factor of cardiovascular events in the patients with chronic kidney disease. High phosphate can induce endothelial cell apoptosis, but the exact mechanism is not clear. This study fills this knowledge gap. Microarray analysis was used

Articles

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service