Skip to Content
Merck
All Photos(3)

Key Documents

SRP0300

Sigma-Aldrich

TGF-β1 Latent human

recombinant, expressed in FreeStyle 293-F cells, ≥98% (SDS-PAGE)

Synonym(s):

LAP, TGFb1, TGFbeta1, latency-associated peptide, transforming growth factor beta-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in FreeStyle 293-F cells

Assay

≥98% (SDS-PAGE)

form

aqueous solution

mol wt

25 kDa

packaging

pkg of 5 μg

manufacturer/tradename

Invitrogen

storage condition

avoid repeated freeze/thaw cycles

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... LTBP1(4052)

General description

Human TGFβ1 (GenBank Accession No. NM_000660), amino acids 30-390 (end) with Cys33Ser mutation and N-terminal His-tag, disulfide-linked homodimers of TGFβ1 LAP and mature TGFβ1, MW= ~75kDa and 25kDa, under non-reducing conditions.
LTBP1 (latent transforming growth factor β binding protein 1) belongs to the LTBP family of proteins. The protein has 18 epidermal growth factor (EGF)-like repeats and four LTBP-like repeats. The gene is mapped to human chromosome 2p22.3.

Application

Useful for cell culture and for the study of signaling and apoptotic pathways.

Biochem/physiol Actions

LTBP1 (latent transforming growth factor β binding protein 1) is an extracellular matrix protein which also controls TGF-β (transforming growth factor β) bioavailability. LTBP1 is mainly responsible for the folding and secretion of TGF-β, thereby adding it to the extracellular matrix. It is a target of HTRA1 (high temperature requirement protein A1), a serine protease involved in cellular signaling.

Physical form

Lyophilized from a 0.2 μM filtered, aqueous solution.

Reconstitution

Reconstitute in sterile PBS with 0.1% BSA. Stability: >6 months at –70°C (as a solid). Store reconstituted protein in aliquots at –80°C for up to 3 months. Avoid freeze/thaw cycles.

Legal Information

FreeStyle is a trademark of Invitrogen Corp.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A suite of novel human spermatozoal RNAs.
Ostermeier GC, et al.
Journal of Andrology, 26, 70-74 (2005)
Association of the small latent transforming growth factor-beta with an eight cysteine repeat of its binding protein LTBP-1.
Saharinen J, et al.
The Embo Journal, 15, 245-253 (1996)
NMR spectroscopic and bioinformatic analyses of the LTBP1 C-terminus reveal a highly dynamic domain organisation.
Robertson IB, et al.
PLoS ONE, 9, e87125-e87125 (2014)
R Derynck et al.
Nature, 316(6030), 701-705 (1985-08-22)
The partial amino-acid sequence of purified human transforming growth factor-beta (TGF-beta) was used to identify a series of cDNA clones encoding the protein. The cDNA sequence indicates that the 112-amino acid monomeric form of the natural TGF-beta homodimer is derived
J S Munger et al.
Kidney international, 51(5), 1376-1382 (1997-05-01)
Transforming growth factor-beta are cytokines with a wide range of biological effects. They play a pathologic role in inflammatory and fibrosing diseases such as nephrosclerosis. TGF-beta s are secreted in a latent form due to noncovalent association with latency associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service