Skip to Content
Merck
All Photos(1)

Key Documents

E9004

Sigma-Aldrich

α-Ecdysone

≥90%

Synonym(s):

2β,3β,14α,22(R),25-Pentahydroxy-7-cholesten-6-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O6
CAS Number:
Molecular Weight:
464.63
Beilstein:
2422986
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

sterility

non-sterile

Assay

≥90%

form

powder

solubility

methanol: 20 mg/mL, clear, colorless to faintly yellow

application(s)

agriculture
environmental

shipped in

ambient

storage temp.

−20°C

SMILES string

[H][C@]12CC[C@]3(C)[C@H](CC[C@@]3(O)C1=CC(=O)[C@]4([H])C[C@@H](O)[C@@H](O)C[C@]24C)[C@H](C)[C@H](O)CCC(C)(C)O

InChI

1S/C27H44O6/c1-15(20(28)8-9-24(2,3)32)16-7-11-27(33)18-12-21(29)19-13-22(30)23(31)14-25(19,4)17(18)6-10-26(16,27)5/h12,15-17,19-20,22-23,28,30-33H,6-11,13-14H2,1-5H3/t15-,16+,17-,19-,20+,22+,23-,25+,26+,27+/m0/s1

InChI key

UPEZCKBFRMILAV-JMZLNJERSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Ecdysone is a phytoecdysteroid, synthesized from cholesterol, and one of the major insect molting hormones. Ecdysone is secreted by the ecdysial glands present in insects.

Application

α-Ecdysone has been used:
  • to study its effects on Drosophila melanogaster′s sexual maturation
  • to study its effects on specification of neurons in Drosphila male-specific neurite
  • as a standard to evaluate the ecdysteroid levels by liquid chromatography-mass spectrometry (LC-MS/MS)

Biochem/physiol Actions

Ecdysone exerts its effects via the ecdysone receptor and regulates larval molting, reproduction, and metamorphosis in insects.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takumi Kayukawa et al.
Developmental biology, 388(1), 48-56 (2014-02-11)
Juvenile hormone (JH) has an ability to repress the precocious metamorphosis of insects during their larval development. Krüppel homolog 1 (Kr-h1) is an early JH-inducible gene that mediates this action of JH; however, the fine hormonal regulation of Kr-h1 and
Kim F Rewitz et al.
Current topics in developmental biology, 103, 1-33 (2013-01-26)
The transition from juvenile to adult is a fundamental process that allows animals to allocate resource toward reproduction after completing a certain amount of growth. In insects, growth to a species-specific target size induces pulses of the steroid hormone ecdysone
Daria Shlyueva et al.
Molecular cell, 54(1), 180-192 (2014-04-02)
Steroid hormones act as important developmental switches, and their nuclear receptors regulate many genes. However, few hormone-dependent enhancers have been characterized, and important aspects of their sequence architecture, cell-type-specific activating and repressing functions, or the regulatory roles of their chromatin
Cecilia Stahl Vieira et al.
Developmental and comparative immunology, 114, 103864-103864 (2020-09-13)
Rhodnius prolixus is an insect vector of two flagellate parasites, Trypanosoma rangeli and Trypanosoma cruzi, the latter being the causative agent of Chagas disease in Latin America. The R. prolixus neuroendocrine system regulates the synthesis of the steroid hormone ecdysone
W Robert Shaw et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(16), 5854-5859 (2014-04-09)
Anopheles gambiae mosquitoes are major African vectors of malaria, a disease that kills more than 600,000 people every year. Given the spread of insecticide resistance in natural mosquito populations, alternative vector control strategies aimed at reducing the reproductive success of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service