Skip to Content
Merck
All Photos(1)

Key Documents

SML3704

Sigma-Aldrich

MRE-269

≥98% (HPLC)

Synonym(s):

2-[4-[(5,6-Diphenyl-2-pyrazinyl)(1-methylethyl)amino]butoxy]-acetic acid, ACT-333679, MRE 269, {4-[(5,6-Diphenylpyrazin-2-yl)(isopropyl)amino]butoxy}acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H29N3O3
CAS Number:
Molecular Weight:
419.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, White to light brown

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

N(C(C)C)(CCCCOCC(=O)O)c1nc(c(nc1)c3ccccc3)c2ccccc2

InChI

1S/C25H29N3O3/c1-19(2)28(15-9-10-16-31-18-23(29)30)22-17-26-24(20-11-5-3-6-12-20)25(27-22)21-13-7-4-8-14-21/h3-8,11-14,17,19H,9-10,15-16,18H2,1-2H3,(H,29,30)

InChI key

OJQMKCBWYCWFPU-UHFFFAOYSA-N

Biochem/physiol Actions

MRE-269 (ACT-333679) is an active form of Selexipag (NS-304). MRE-269 is a potent and highly selective IP receptor agonist (selective PGI2 receptor agonist).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidekazu Maruyama et al.
Life sciences, 315, 121372-121372 (2023-01-08)
Upregulated p38MAPK signaling is implicated in the accelerated proliferation of pulmonary artery smooth muscle cells (PA-SMCs) and the pathogenesis of pulmonary artery remodeling observed in pulmonary arterial hypertension (PAH). Previously, we reported that after endothelin-1 (ET-1) pretreatment, bone morphogenetic protein
Samuel N Baldwin et al.
British journal of pharmacology, 179(7), 1338-1352 (2021-11-13)
Prostacyclin mimetics express potent vasoactive effects via prostanoid receptors that are not unequivocally defined, as to date no study has considered sex as a factor. The aim of this study was to determine the contribution of IP and EP3 prostanoid
Keith Morrison et al.
The Journal of pharmacology and experimental therapeutics, 335(1), 249-255 (2010-07-28)
Selexipag [2-{4-[(5,6-diphenylpyrazin-2-yl)(isopropyl)amino]butoxy}-N-(methylsulfonyl)acetamide] is an orally available prostacyclin (PGI(2)) receptor (IP receptor) agonist that is chemically distinct from PGI(2) and is in clinical development for the treatment of pulmonary arterial hypertension. Selexipag is highly selective for the human IP receptor in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service