Skip to Content
Merck
All Photos(1)

Key Documents

SML2872

Sigma-Aldrich

EIDD-1931

≥98% (HPLC)

Synonym(s):

(oh)4Cyd, 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-(hydroxyamino)pyrimidin-2(1H)-one, 4-N-HO-Cyd, BHC, EIDD 1931, EIDD 2801 metabolite, EIDD-2801 metabolite, EIDD1931, EIDD2801 metabolite, N4-Hydroxycytidine, N4-OHcyd, N4OHcyd, NHC, beta-D-N4-Hydroxycytidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O6
CAS Number:
Molecular Weight:
259.22
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[n]2([c](nc(cc2)NO)=O)[C@@H]1O[C@@H]([C@H]([C@H]1O)O)CO

InChI

1S/C9H13N3O6/c13-3-4-6(14)7(15)8(18-4)12-2-1-5(11-17)10-9(12)16/h1-2,4,6-8,13-15,17H,3H2,(H,10,11,16)/t4-,6-,7-,8-/m1/s1

InChI key

XCUAIINAJCDIPM-XVFCMESISA-N

Biochem/physiol Actions

Originally recognized as a powerful mutagen that preferentially induces AT-to-GC transition in microorganisms, EIDD-1931 (NHC, N4-hydroxycytidine) is an orally available ribonucleoside analog with known anti-viral activity against SARS-CoV, SARS-CoV-2, respiratory syncytial virus (RSV), influenza A & B viruses (IAV & IBV), HCV, pestivirus, bovine viral diarrhoea virus (BVDV), Ebola virus (EBOV), Chikungunya virus (CHIKV), venezuelan equine encephalitis virus (VEEV). NHC effectively reduces RSV and IAVs replication in mice, and exhibts efficacy in a guinea pig IAV transmission model in vivo (100 & 400 mg/kg b.i.d. p.o.). Metabolic studies reveal species-dependent plasma stability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George R Painter et al.
Antiviral research, 171, 104597-104597 (2019-09-09)
The New World alphaviruses Venezuelan, Eastern, and Western equine encephalitis viruses (VEEV, EEEV and WEEV, respectively) commonly cause a febrile disease that can progress to meningoencephalitis, resulting in significant morbidity and mortality. To address the need for a therapeutic agent
Maria L Agostini et al.
Journal of virology, 93(24) (2019-10-04)
Coronaviruses (CoVs) have emerged from animal reservoirs to cause severe and lethal disease in humans, but there are currently no FDA-approved antivirals to treat the infections. One class of antiviral compounds, nucleoside analogues, mimics naturally occurring nucleosides to inhibit viral
Maryam Ehteshami et al.
Antimicrobial agents and chemotherapy, 61(4) (2017-02-01)
Chikungunya virus (CHIKV) represents a reemerging global threat to human health. Recent outbreaks across Asia, Europe, Africa, and the Caribbean have prompted renewed scientific interest in this mosquito-borne alphavirus. There are currently no vaccines against CHIKV, and treatment has been
Mart Toots et al.
Translational research : the journal of laboratory and clinical medicine, 218, 16-28 (2020-01-17)
Seasonal influenza viruses cause major morbidity and mortality worldwide, threatening in particular older adults and the immunocompromised. Two classes of influenza therapeutics dominate current disease management, but both are compromised by pre-existing or rapidly emerging viral resistance. We have recently
Mart Toots et al.
Science translational medicine, 11(515) (2019-10-28)
Influenza viruses constitute a major health threat and economic burden globally, frequently exacerbated by preexisting or rapidly emerging resistance to antiviral therapeutics. To address the unmet need of improved influenza therapy, we have created EIDD-2801, an isopropylester prodrug of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service