Skip to Content
Merck
All Photos(1)

Key Documents

SML2822

Sigma-Aldrich

AZ12601011

≥98% (HPLC)

Synonym(s):

1-(2-Pyridin-2-yl-6,7dihydro-5H-cyclopenta[e]pyrimidin-4-yl)pyrrolo[3,2-c]pyridine, 2-(2-Pyridinyl)-4-(1H-pyrrolo[3,2-c]pyridin-1-yl)-6,7-dihydro-5H-cyclopenta[d]pyrimidine, AZ 12601011, AZ-12601011

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15N5
Molecular Weight:
313.36
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

AZ12601011 is an orally active, subtype-selective TGF-β type I receptors active site inhibitor (active-site affinity: ALK4/5/6 Kd = 2.6/2.9/42 nM, ALK1/2/3 Kd = 40/15/40 μM) that prevents ALK4/5/7-mediated Smad2, but not ALK1/2/3/6-mediated Smad1, phosphorylation (10 μM; NIH3T3 expressing respective constitutively active receptors). AZ12601011 inhibits 4T1 murine mammary carcinoma growth in cultures (IC50 = 400 nM) and in vivo (50 mg/kg bid. po. mice). AZ12601011 also causes heart valve lesions and physeal dysplasia in rats (150 mg/kg/day po. for 7 days), consistent with a critical role of ALK5 in maintaining the integrity of heart valve and physis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas G Bird et al.
Science translational medicine, 10(454) (2018-08-17)
Liver injury results in rapid regeneration through hepatocyte proliferation and hypertrophy. However, after acute severe injury, such as acetaminophen poisoning, effective regeneration may fail. We investigated how senescence may underlie this regenerative failure. In human acute liver disease, and murine
Mark J Anderton et al.
Toxicologic pathology, 39(6), 916-924 (2011-08-24)
Aberrant signaling by transforming growth factor-β (TGF-β) and its type I (ALK5) receptor has been implicated in a number of human diseases and this pathway is considered a potential target for therapeutic intervention. Transforming growth factor-β signaling via ALK5 plays
Lindsay C Spender et al.
Molecular pharmacology, 95(2), 222-234 (2018-11-22)
The transforming growth factor β (TGFβ) superfamily includes TGFβ, activins, inhibins, and bone morphogenetic proteins (BMPs). These extracellular ligands have essential roles in normal tissue homeostasis by coordinately regulating cell proliferation, differentiation, and migration. Aberrant signaling of superfamily members, however
Rene Jackstadt et al.
Cancer cell, 36(3), 319-336 (2019-09-19)
The metastatic process of colorectal cancer (CRC) is not fully understood and effective therapies are lacking. We show that activation of NOTCH1 signaling in the murine intestinal epithelium leads to highly penetrant metastasis (100% metastasis; with >80% liver metastases) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service