Skip to Content
Merck
All Photos(1)

Documents

16-240

Sigma-Aldrich

Normal Mouse IgG, Alexa Fluor 488 conjugate

Normal Mouse IgG Polyclonal Antibody control validated for use in Immunofluorescence.

Synonym(s):

Normal Mouse IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

clone

polyclonal

manufacturer/tradename

Upstate®

technique(s)

immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Application

Normal Mouse IgG Polyclonal Antibody control validated for use in Immunofluorescence.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Physical form

Fractionation and ion exchange chromatography

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hannah B Gordon et al.
Development (Cambridge, England), 145(22) (2018-10-20)
Establishment of precise three-dimensional tissue structure is vital for organ function. In the visual system, optic fissure and stalk morphogenesis is a crucial yet poorly understood process, disruptions of which can lead to coloboma, a birth defect causing visual impairment.
Jian Luo et al.
International journal of molecular sciences, 23(14) (2022-07-28)
Microsporidia are obligate intracellular parasites that infect a wide variety of hosts ranging from invertebrates to vertebrates. These parasites have evolved strategies to directly hijack host mitochondria for manipulating host metabolism and immunity. However, the mechanism of microsporidia interacting with
Patricia R Braun et al.
Translational psychiatry, 9(1), 47-47 (2019-02-02)
Differential DNA methylation in the brain is associated with many psychiatric diseases, but access to brain tissues is essentially limited to postmortem samples. The use of surrogate tissues has become common in identifying methylation changes associated with psychiatric disease. In
Matthew J Crow et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 79(1), 57-65 (2010-12-25)
Fluorescence-based flow cytometry measures multiple cellular characteristics, including levels of receptor expression, by assessing the fluorescence intensity from a population of cells whose cell surface receptors are bound by a fluorescently labeled antibody or ligand for that receptor. Functionalized noble
Hong Seok Choi et al.
Frontiers in oncology, 14, 1376831-1376831 (2024-05-22)
Cancer metastasis is dependent on cell migration. Several mechanisms, including epithelial-to-mesenchymal transition (EMT) and actin fiber formation, could be involved in cancer cell migration. As a downstream effector of the Hippo signaling pathway, transcriptional coactivator with PDZ-binding motif (TAZ) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service