Skip to Content
Merck
All Photos(1)

Documents

SML2270

Sigma-Aldrich

FRAX486

≥98% (HPLC)

Synonym(s):

6-(2,4-Dichlorophenyl)-8-ethyl-2-(3-fluoro-4-(piperazin-1-yl)phenylamino)pyrido[2,3-d] pyrimidin-7(8H)-one, 6-(2,4-Dichlorophenyl)-8-ethyl-2-[[3-fluoro-4-(1-piperazinyl)phenyl]amino]pyrido[2,3-d]pyrimidin-7(8H)-one, FRAX 486, FRAX-486

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23Cl2FN6O
CAS Number:
Molecular Weight:
513.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

FC1=CC(NC2=NC(N3CC)=C(C=N2)C=C(C4=CC=C(C=C4Cl)Cl)C3=O)=CC=C1N5CCNCC5

InChI

1S/C25H23Cl2FN6O/c1-2-34-23-15(11-19(24(34)35)18-5-3-16(26)12-20(18)27)14-30-25(32-23)31-17-4-6-22(21(28)13-17)33-9-7-29-8-10-33/h3-6,11-14,29H,2,7-10H2,1H3,(H,30,31,32)

InChI key

DHKFOIHIUYFSOF-UHFFFAOYSA-N

Biochem/physiol Actions

FRAX486 is a potent, group I-selective p21-activated kinase (PAK) inhibitor (PAK1/2/3 IC50 = 8.25/39.5/55.3 nM; group II PAK4 IC50 = 779 nM). FRAX486 effectively inhibits PAK-mediated PKD1 Ser203 phosphorylation and PAK1/2 Ser144/141 autophosphorylation upon angiotensin II (ANGII) stimulation of rat IEC-18 cells (IC50 ∼0.5 μM) and exhibits in vivo efficacy in rescuing the autism-like phenotypes among Fmr1 knockout fragile syndrome (FXS) mice (20 mg/kg s.c.) as well as in ameliorating schizophrenia-associated dendritic spine deterioration among Disc1 knockdown mice (10 μg/g or 10 mg/kg i.p.) with good pharmacokinetic properties and blood–brain barrier (BBB) permeability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akiko Hayashi-Takagi et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(17), 6461-6466 (2014-04-08)
Drug discovery in psychiatry has been limited to chemical modifications of compounds originally discovered serendipitously. Therefore, more mechanism-oriented strategies of drug discovery for mental disorders are awaited. Schizophrenia is a devastating mental disorder with synaptic disconnectivity involved in its pathophysiology.
Alexander Pyronneau et al.
Science signaling, 10(504) (2017-11-09)
Fragile X syndrome (FXS) is the most common inherited cause of intellectual disabilities and a leading cause of autism. FXS is caused by a trinucleotide expansion in the gene FMR1 on the X chromosome. The neuroanatomical hallmark of FXS is
Jen-Kuan Chang et al.
The Journal of biological chemistry, 292(23), 9523-9539 (2017-04-15)
Although PKC-mediated phosphorylation of protein kinase D1 (PKD1) has been extensively characterized, little is known about PKD1 regulation by other upstream kinases. Here we report that stimulation of epithelial or fibroblastic cells with G protein-coupled receptor agonists, including angiotensin II
Joju George et al.
eLife, 4, e06327-e06327 (2015-04-18)
Precise regulation of the dendritic spine actin cytoskeleton is critical for neurodevelopment and neuronal plasticity, but how neurons spatially control actin dynamics is not well defined. Here, we identify direct palmitoylation of the actin regulator LIM kinase-1 (LIMK1) as a
Alexei S Karpov et al.
ACS medicinal chemistry letters, 6(7), 776-781 (2015-07-21)
The discovery of inhibitors targeting novel allosteric kinase sites is very challenging. Such compounds, however, once identified could offer exquisite levels of selectivity across the kinome. Herein we report our structure-based optimization strategy of a dibenzodiazepine hit 1, discovered in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service