Skip to Content
Merck
All Photos(3)

Key Documents

PZ0162

Sigma-Aldrich

PD 0325901

≥98% (HPLC)

Synonym(s):

N-[(2R)-2,3-Dihydroxypropoxy]-3,4-difluoro-2-[(2-fluoro-4-iodophenyl)amino]-benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14F3IN2O4
CAS Number:
Molecular Weight:
482.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -1.8 to -3.5°, c = 1 mg/mL in methanol

color

white to off-white

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=C(NC2=CC=C(I)C=C2F)C(C(NOC[C@H](O)CO)=O)=CC=C1F

InChI

1S/C16H14F3IN2O4/c17-11-3-2-10(16(25)22-26-7-9(24)6-23)15(14(11)19)21-13-4-1-8(20)5-12(13)18/h1-5,9,21,23-24H,6-7H2,(H,22,25)/t9-/m1/s1

InChI key

SUDAHWBOROXANE-SECBINFHSA-N

Gene Information

Application

PD 0325901 has been used as an inhibitor in extracellular signal-regulated kinase (ERK) inhibition assay in primed induced pluripotent stem cells, RKO colorectal cancer cell line and human embryonic stem cells(hESCs).

Biochem/physiol Actions

PD 0325901 by inhibiting mitogen-activated protein kinases (MAPKs) elicits growth-inhibitory and antiangiogenic effects on glioblastoma and melanoma tumor progression.
PD 0325901 is a potent MKK1 (MEK1) and MKK2 (MEK2) inhibitor. The Ki is 1.1 and 0.79 nM for MEK1 and MEK2, respectively. PD 0325901 was inactive against a panel of 27 other kinases. PD 0325901 inhibited C26 tumor pERK by 75% when dosed at 25 mg/kg in mice.

Features and Benefits

This compound is featured on the MAPKKKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of glioblastoma dispersal by the MEK inhibitor PD0325901
Shannon S, et al.
BMC Cancer, 17(1), 121-121 (2017)
Growth-inhibitory and antiangiogenic activity of the MEK inhibitor PD0325901 in malignant melanoma with or without BRAF mutations
Ciuffreda L, et al.
Neoplasia, 11(8), 720-W6-720-W6 (2009)
Bin Zhang et al.
The Journal of clinical investigation, 126(3), 975-991 (2016-02-16)
Chronic myelogenous leukemia (CML) results from transformation of a long-term hematopoietic stem cell (LTHSC) by expression of the BCR-ABL fusion gene. However, BCR-ABL-expressing LTHSCs are heterogeneous in their capacity as leukemic stem cells (LSCs). Although discrepancies in proliferative, self-renewal, and
Madeleine Linneberg-Agerholm et al.
Development (Cambridge, England), 146(24) (2019-11-20)
Embryonic stem cells (ESCs) exist in at least two states that transcriptionally resemble different stages of embryonic development. Naïve ESCs resemble peri-implantation stages and primed ESCs the pre-gastrulation epiblast. In mouse, primed ESCs give rise to definitive endoderm in response
Feeder-Free Derivation of Naive Human Pluripotent Stem Cells
Ward E, et al.
Stem Cells and Development, 26(15), 1087-1089 (2017)

Articles

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service