Skip to Content
Merck
All Photos(1)

Key Documents

SML1933

Sigma-Aldrich

TP-472N

≥98% (HPLC)

Synonym(s):

1-[8-[2-(Cyclopropylmethoxy)phenyl]pyrrolo[1,2-a]pyrimidin-6-yl]-ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N2O2
CAS Number:
Molecular Weight:
306.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C)C1=CC(C2=CC=CC=C2OCC3CC3)=C4N=CC=CN41

InChI key

IDQWBZAMTQTECP-UHFFFAOYSA-N

Biochem/physiol Actions

TP-472N is a negative control probe for TP-472. TP-472 is a selective probe for bromodomain-containing proteins BRD9 and BRD7. The BRD9/7 probe was developed out of a collaborative effort by Takeda and the SGC, and is of a different chemotype from BRD9/7 probes previously available. TP-472 has a KD value of 33nM for BRD9 and 340nM for BRD7 by ITC, is >30 fold selective for BRD9 over all other bromodomain family members except BRD7, and is suitable for in vivo applications. The negative control TP-472N is inactive against other bromodomains (>20μM against BRD9). For full characterization details of TP-472 and TP-472N, please visit the TP-472 probe summary on the Structural Genomics Consortium (SGC) website.

TP-472, the active BRD9/7 probe, is available from Sigma. To learn more about and purchase TP-472, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

TP-472N is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service