Skip to Content
Merck
All Photos(1)

Key Documents

SML1402

Sigma-Aldrich

TCID

≥98% (HPLC)

Synonym(s):

4,5,6,7-Tetrachloro-1,3-indandione, 4,5,6,7-Tetrachloro-1H-indene-1,3(2H)-dione, CID 2729042, UCH-L3 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H2Cl4O2
CAS Number:
Molecular Weight:
283.92
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1CC(C2=C(Cl)C(Cl)=C(Cl)C(Cl)=C21)=O

InChI

1S/C9H2Cl4O2/c10-6-4-2(14)1-3(15)5(4)7(11)9(13)8(6)12/h1H2

InChI key

IDLAOWFFKWRNHB-UHFFFAOYSA-N

Biochem/physiol Actions

TCID is a cell penetrant potent and specific inhibitor of UCH-L3 (Ubiquitin carboxyl-terminal hydrolase isozyme L3). TCID is used to distinguish between UCH-L1 and UCH-L3 activities in cells.
TCID is a cell penetrant potent and specific inhibitor of UCH-L3.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anurag Rathore et al.
Scientific reports, 10(1), 5350-5350 (2020-03-27)
The major barrier to a HIV-1 cure is the persistence of latent genomes despite treatment with antiretrovirals. To investigate host factors which promote HIV-1 latency, we conducted a genome-wide functional knockout screen using CRISPR-Cas9 in a HIV-1 latency cell line

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service