Skip to Content
Merck
All Photos(2)

Key Documents

684163

Sigma-Aldrich

Indan-1,2-dione

97%

Synonym(s):

1H-Indene-1,2(3H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

117-124 °C

functional group

ketone

SMILES string

O=C1Cc2ccccc2C1=O

InChI

1S/C9H6O2/c10-8-5-6-3-1-2-4-7(6)9(8)11/h1-4H,5H2

InChI key

WFFZGYRTVIPBFN-UHFFFAOYSA-N

Application

Indan-1,2-dione is a latent fingerprint reagent widely used in forensic chemistry. It can also be used to synthesize polycyclic arene and heteroarene systems containing five membered rings.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bent N-Heteroarenes.
Hahn S, et al.
The Journal of Organic Chemistry, 81(18), 8485-8494 (2016)
A Combined Experimental and Theoretical Study of the Ammonium Bifluoride Catalyzed Regioselective Synthesis of Quinoxalines and Pyrido[2,3-b]pyrazines.
Lassagne F, et al.
Synthesis, 47, 2680-2689 (2015)
Optimisation and evaluation of 1,2-indanedione for use as a fingermark reagent and its application to real samples.
Wallace-Kunkel C, et al.
Forensic Science International, 168(1), 14-26 (2007)
Patrick Fritz et al.
Forensic science international, 257, 20-28 (2015-08-19)
Dimethylaminocinnamaldehyde was re-evaluated as a wet contact reagent for the treatment of latent fingermarks on porous substrates. A new formulation (consisting of 0.028 g p-dimethylaminocinnamaldehyde, 0.84 mL glacial acetic acid, 6.2 mL ethyl acetate and 0.993 L 40-60 °C petroleum
Chemical development of latent fingerprints: 1, 2-indanedione has come of age.
Wiesner S, et al.
Journal of Forensic Sciences, 46(5), 1082-1084 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service