Skip to Content
Merck
All Photos(1)

Key Documents

SML1162

Sigma-Aldrich

5-Fluoro-1-propargyl-uracil

≥98% (HPLC)

Synonym(s):

1-Propargyl-5-fluorouracil, 5-Fluoro-1-(2-propyn-1-yl)-2,4(1H,3H)-pyrimidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5FN2O2
CAS Number:
Molecular Weight:
168.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C1C(F)=CN(CC#C)C(N1)=O

InChI

1S/C7H5FN2O2/c1-2-3-10-4-5(8)6(11)9-7(10)12/h1,4H,3H2,(H,9,11,12)

InChI key

GMCOGJQJFDQZCP-UHFFFAOYSA-N

Biochem/physiol Actions

5-Fluoro-1-propargyl-uracil is a biologically inert precursor, a prodrug that can be bioorthogonally activated to 5-fluorouracil iside a tumor that has a previously implanted palladium catalyst. Preliminary tests with pancreatic and colorectal cancer cells resulted in the death of than 80% of the malignant cells.
5-Fluoro-1-propargyl-uracil is a biologically inert precursor, a prodrug that can be bioorthogonally activated to 5-fluorouracil.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service