Skip to Content
Merck
All Photos(2)

Key Documents

722332

Sigma-Aldrich

6-Amino-7-deazapurine

97%

Synonym(s):

4-Amino-7H-pyrrolo[2,3-d]pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4
CAS Number:
Molecular Weight:
134.14
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Assay

97%

form

solid

mp

257-262 °C

SMILES string

Nc1ncnc2[nH]ccc12

InChI

1S/C6H6N4/c7-5-4-1-2-8-6(4)10-3-9-5/h1-3H,(H3,7,8,9,10)

InChI key

PEHVGBZKEYRQSX-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Ono et al.
Nucleic acids research, 12(23), 8939-8949 (1984-12-11)
Deoxydecanucleotides having a recognition sequence of Bgl II and Sau 3AI, and their 7-deazaadenine analogs were synthesized. The decanucleotides containing 7-deazaadenine in place of adenine were partially resistant to the hydrolysis by Sau 3AI and strongly resistant to that by
Munmun Maiti et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(3), 857-868 (2011-12-17)
Nucleoside phosphoramidates (NPs) are a class of nucleotide analogues that has been developed as potential antiviral/antitumor prodrugs. Recently, we have shown that some amino acid nucleoside phosphoramidates (aaNPs) can act as substrates for viral polymerases like HIV-1 RT. Herein, we report
Pavel Kielkowski et al.
The Journal of organic chemistry, 76(9), 3457-3462 (2011-03-24)
(Cytosin-5-yl)ethynyl derivatives of pyrimidine and 7-deazaadenine 2-deoxyribonucleosides and nucleoside triphosphates (dNTPs) were prepared in one step by the aqueous Sonogashira coupling of unprotected halogenated nucleos(t)ides with 5-ethynylcytosine. The modified dNTPs were good substrates for DNA polymerases suitable for primer extension
Akimitsu Okamoto et al.
Bioorganic & medicinal chemistry letters, 12(1), 97-99 (2001-12-12)
2-Amino-7-deazaadenine ((AD)A) was incorporated into oligodeoxynucleotides (ODN) and their base-pairing properties with natural nucleobases were investigated. In melting temperature (T(m)) experiments, the duplex containing an (AD)A/C base pair showed a high stability comparable to that containing (AD)A/T base pair. Destabilization
A I Murchie et al.
The EMBO journal, 13(4), 993-1001 (1994-02-15)
Telomeres are required for eukaryotic chromosome stability. They consist of regularly repeating guanine-rich sequences, with a single-stranded 3' terminus. Such sequences have been demonstrated to have the propensity to adopt four-stranded structures based on a tetrad of guanine bases. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service