Skip to Content
Merck
All Photos(1)

Key Documents

SML1044

Sigma-Aldrich

Zosuquidar hydrochloride

≥98% (HPLC), powder, p-Glycoprotein inhibitor

Synonym(s):

(2R)-Anti-5-[3-[4-(10,11-difluoromethanodibenzosuber-5-yl)piperazin-1-yl]-2-hydroxypropoxy]quinoline hydrochloride, LY-335979, RS-33295-198

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H31F2N3O2 · xHCl
CAS Number:
Molecular Weight:
527.60 (free base basis)
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

product name

Zosuquidar hydrochloride, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 4 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

O[C@@H](COC1=C(C=CC=N2)C2=CC=C1)CN(CC3)CCN3[C@@H]4C5=C(C=CC=C5)[C@@H]6[C@@H](C6(F)F)C7=C4C=CC=C7.Cl

InChI

1S/C32H31F2N3O2.ClH/c33-32(34)29-22-7-1-3-9-24(22)31(25-10-4-2-8-23(25)30(29)32)37-17-15-36(16-18-37)19-21(38)20-39-28-13-5-12-27-26(28)11-6-14-35-27;/h1-14,21,29-31,38H,15-20H2;1H/t21-,29-,30+,31-;/m1./s1

InChI key

VQJFFWJUYDGTQZ-FCNWNIDBSA-N

Biochem/physiol Actions

Zosuqidar is a potent inhibitor of P-glycoprotein (P-gp, MDR1) a transporter protein that is a key modulator of cellular drug efflux. Zosuquidar sensitizes AML, and other cancer cell lines to cytotoxic drugs.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yoko Nagaya et al.
Drug metabolism and disposition: the biological fate of chemicals, 48(11), 1183-1190 (2020-08-31)
Estimation of unbound drug concentration in the brain (Cu,brain) is an essential part of central nervous system (CNS) drug development. As a surrogate for Cu,brain in humans and nonhuman primates, drug concentration in cerebrospinal fluid (CCSF) collected by lumbar puncture
J Vérité et al.
Journal of neuroinflammation, 15(1), 182-182 (2018-06-15)
Alzheimer's disease is widely described since the discovery of histopathological lesions in Mrs. Auguste Deter in 1906. However to date, there is no effective treatment to deal with the many cellular and molecular alterations. The complexity is even higher with
Kamil Nosol et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(42), 26245-26253 (2020-10-07)
ABCB1 detoxifies cells by exporting diverse xenobiotic compounds, thereby limiting drug disposition and contributing to multidrug resistance in cancer cells. Multiple small-molecule inhibitors and inhibitory antibodies have been developed for therapeutic applications, but the structural basis of their activity is
Shehab Eid et al.
International journal of molecular sciences, 23(23) (2022-12-12)
Several strands of investigation have established that a reduction in the levels of the cellular prion protein (PrPC) is a promising avenue for the treatment of prion diseases. We recently described an indirect approach for reducing PrPC levels that targets
Nienke R Wevers et al.
Fluids and barriers of the CNS, 18(1), 59-59 (2021-12-16)
In ischemic stroke, the function of the cerebral vasculature is impaired. This vascular structure is formed by the so-called neurovascular unit (NVU). A better understanding of the mechanisms involved in NVU dysfunction and recovery may lead to new insights for

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service