Skip to Content
Merck
All Photos(1)

Key Documents

SML0896

Sigma-Aldrich

MCC-555

≥98% (HPLC)

Synonym(s):

5-[[6-[(2-Fluorophenyl)methoxy]-2-naphthalenyl]methyl]-2,4-thiazolidinedione, Isaglitazone, Netoglitazone, RWJ 241947

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16FNO3S
CAS Number:
Molecular Weight:
381.42
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C21H16FNO3S/c22-18-4-2-1-3-16(18)12-26-17-8-7-14-9-13(5-6-15(14)11-17)10-19-20(24)23-21(25)27-19/h1-9,11,19H,10,12H2,(H,23,24,25)

InChI key

PKWDZWYVIHVNKS-UHFFFAOYSA-N

Biochem/physiol Actions

MCC-555 is a thiazolidinedione class anti-diabetic compound. In rodent models, MCC-555 attenuates the development of diabetes, maintains B-cell function and improves insulin sensitivity. The compound MCC-555 also inhibits proliferation of several cancer cell lines and reduces tumor growth in xenograft models.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ulrich et al.
Experimental cell research, 310(1), 196-204 (2005-08-23)
Histone deacetylase-inhibitors affect growth and differentiation of intestinal epithelial cells by inducing expression of several transcription factors, e.g. Peroxisome proliferator-activated receptor gamma (PPARgamma) or vitamin D receptor (VDR). While activation of VDR by butyrate mainly seems to be responsible for
L Pickavance et al.
British journal of pharmacology, 125(4), 767-770 (1998-12-01)
1. Young (6-week-old) pre-diabetic Zucker Diabetic Fatty (ZDF) rats displaying impaired glucose tolerance (IGT), moderate hyperglycaemia and hyperinsulinaemia were treated with the novel thiazolidinedione, MCC-555, for 28 days, during which time beta-cell failure and progression to overt diabetes occurs. 2.
Kyung-Won Min et al.
Toxicology and applied pharmacology, 263(2), 225-232 (2012-07-04)
MCC-555 is a novel PPARα/γ dual ligand of the thiazolidinedione class and was recently developed as an anti-diabetic drug with unique properties. MCC-555 also has anti-proliferative activity through growth inhibition and apoptosis induction in several cancer cell types. Our group
Kiyoshi Yamaguchi et al.
Molecular cancer therapeutics, 5(5), 1352-1361 (2006-05-30)
Apoptosis and/or differentiation induction caused by the peroxisome proliferator-activated receptor gamma (PPARgamma) ligand is a promising approach to cancer therapy. The thiazolidinedione derivative MCC-555 has an apoptotic activity in human colorectal cancer cells, accompanied by up-regulation of a proapoptotic nonsteroidal
M J Reginato et al.
The Journal of biological chemistry, 273(49), 32679-32684 (1998-11-26)
Thiazolidinediones (TZDs) constitute an exciting new class of antidiabetic compounds, which function as activating ligands for peroxisome proliferator-activated receptor gamma (PPARgamma). Until now, there has been an excellent correlation between in vivo hypoglycemic potency and in vitro binding and activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service