Skip to Content
Merck
All Photos(2)

Key Documents

435244

Sigma-Aldrich

Cellulose

colloidal, microcrystalline

Synonym(s):

Cellulose powder, Cotton linters

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

colloidal, microcrystalline

contains

10.0-20.0% sodium carboxymethyl cellulose as stabilizer

particle size

+65-+325 mesh

viscosity

50-150 cP, 2.6 wt. % in H2O, Brookfield RTV, spindle #1 (20 rpm)(lit.)

bulk density

0.6 g/mL (25°C)

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bulking agent, opacifier, anti-caking agent, extrusion aid and stabilizer for foams and emulsions.
High purity cellulose powders for partition chromatography.

Features and Benefits

Amorphous regions are hydrolyzed leaving crystalline microfibrils. Forms thixotropic gels, good thermal stability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Wang et al.
Chemical Society reviews, 41(4), 1519-1537 (2012-01-24)
Utilization of natural polymers has attracted increasing attention because of the consumption and over-exploitation of non-renewable resources, such as coal and oil. The development of green processing of cellulose, the most abundant biorenewable material on Earth, is urgent from the
Edmond Lam et al.
Trends in biotechnology, 30(5), 283-290 (2012-03-13)
Nanocrystalline cellulose (NCC), a rod-shaped nanoscale material with exceptional strength and physicochemical properties, can be prepared from inexpensive renewable biomass. Besides its potential use as a reinforcing agent for industrial biocomposites, pristine NCC exhibits low toxicity and poses no serious
Heather E McFarlane et al.
Annual review of plant biology, 65, 69-94 (2014-03-04)
Plant stature and development are governed by cell proliferation and directed cell growth. These parameters are determined largely by cell wall characteristics. Cellulose microfibrils, composed of hydrogen-bonded β-1,4 glucans, are key components for anisotropic growth in plants. Cellulose is synthesized
Orly Salama-Alber et al.
The Journal of biological chemistry, 288(23), 16827-16838 (2013-04-13)
The rumen bacterium Ruminococcus flavefaciens produces a highly organized multienzyme cellulosome complex that plays a key role in the degradation of plant cell wall polysaccharides, notably cellulose. The R. flavefaciens cellulosomal system is anchored to the bacterial cell wall through
Akihiko Nakamura et al.
The Journal of biological chemistry, 288(19), 13503-13510 (2013-03-28)
Mutation of Trp-40 in the Cel7A cellobiohydrolase from Trichoderma reesei (TrCel7A) causes a loss of crystalline cellulose-degrading ability. Mutant W40A showed reduced specific activity for crystalline cellulose and diffused the cellulose chain from the entrance of the active site tunnel.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service