Skip to Content
Merck
All Photos(1)

Key Documents

SML0853

Sigma-Aldrich

SB 699551 dihydrochloride

≥98% (HPLC)

Synonym(s):

N-[2-(Dimethylamino)ethyl]-N-[[4′-[[(2-phenylethyl)amino]methyl][1,1′-biphenyl]-4-yl]methyl]-cyclopentanepropanamide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H45N3O · 2HCl
CAS Number:
Molecular Weight:
584.66
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Cl.Cl.N(CCN(Cc2ccc(cc2)c3ccc(cc3)CNCCc4ccccc4)C(=O)CCC1CCCC1)(C)C

InChI

1S/C34H45N3O.2ClH/c1-36(2)24-25-37(34(38)21-16-28-10-6-7-11-28)27-31-14-19-33(20-15-31)32-17-12-30(13-18-32)26-35-23-22-29-8-4-3-5-9-29;;/h3-5,8-9,12-15,17-20,28,35H,6-7,10-11,16,21-27H2,1-2H3;2*1H

InChI key

QJMKBIHLMPTYTI-UHFFFAOYSA-N

Application

SB 699551 dihydrochloride has been used to identify the antagonism of 5-HT2AR in cell culture-derived hepatitis C virus (HCVcc).

Biochem/physiol Actions

SB 699551 dihydrochloride is also known as (3-cyclopentyl-N-[2-(dimethylamino)ethyl]-N-[(4′-{[(2-phenylethyl)amino]methyl}-4-biphenylyl)methyl]propanamide dihydrochloride). It is considered as a useful tool to explain the physiological roles of the 5-ht5A receptor.
SB-699551 is 5-HT5A receptor antagonist.
SB-699551 is a 5-HT5A receptor antagonist, (pKi = 8.3) with 100-fold selectivity over other 5-HT receptor populations. SB-699551 completely blocks 5-HT-induced [35S]GTPγS membrane binding in cells expressing recombinant guinea pig 5-HT-5A.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5-ht5A receptors as a therapeutic target
Thomas, David R
Pharmacology & Therapeutics, 111(3), 707-714 (2006)
Identification of serotonin 2A receptor as a novel HCV entry factor by a chemical biology strategy
Cao, et al.
Protein & cell, 1-18 (2018)
Lin Cao et al.
Protein & cell, 10(3), 178-195 (2018-03-16)
Hepatitis C virus (HCV) is a leading cause of liver disease worldwide. Although several HCV protease/polymerase inhibitors were recently approved by U.S. FDA, the combination of antivirals targeting multiple processes of HCV lifecycle would optimize anti-HCV therapy and against potential

Articles

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service