Skip to Content
Merck
All Photos(1)

Key Documents

M6940

Sigma-Aldrich

Modafinil

≥98% (HPLC)

Synonym(s):

2-[(Diphenylmethyl)sulfinyl]­acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15NO2S
CAS Number:
Molecular Weight:
273.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

drug control

USDEA Schedule IV

color

white

solubility

DMSO: >10 mg/mL

originator

Teva

SMILES string

NC(=O)CS(=O)C(c1ccccc1)c2ccccc2

InChI

1S/C15H15NO2S/c16-14(17)11-19(18)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H2,16,17)

InChI key

YFGHCGITMMYXAQ-UHFFFAOYSA-N

Gene Information

human ... SLC6A3(6531)
rat ... Slc6a3(24898)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Modafinil is a central nervous system vigilance promoting agent, which posseses neuroprotective properties.

Features and Benefits

This compound was developed by Teva. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kousuke Okimura et al.
Scientific reports, 11(1), 1843-1843 (2021-01-21)
At high latitudes, approximately 10% of people suffer from depression during the winter season, a phenomenon known as seasonal affective disorder (SAD). Shortened photoperiod and/or light intensity during winter season are risk factors for SAD, and bright light therapy is
Derek Dunn et al.
Chemical biology & drug design, 81(3), 433-435 (2012-12-05)
Utilizing atypical wake-promoting agent modafinil (inactive in both rH(3) and hH(3) binding assays) as a launching pad, a series of sulfinyl- and sulfone-derived H(3) receptor inverse agonists were developed. Brain-permeable compound 27, a potent member of the series displayed excellent
Trends in on-label and off-label modafinil use in a nationally representative sample.
Renée A Peñaloza et al.
JAMA internal medicine, 173(8), 704-706 (2013-04-05)
Mariana Raineri et al.
PloS one, 7(10), e46599-e46599 (2012-10-12)
Methamphetamine is a drug of abuse that can cause neurotoxic damage in humans and animals. Modafinil, a wake-promoting compound approved for the treatment of sleeping disorders, is being prescribed off label for the treatment of methamphetamine dependence. The aim of
Hypersomnia due to bilateral thalamic lesions: unexpected response to Modafinil.
I Zavalko et al.
European journal of neurology, 19(11), e125-e126 (2012-08-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service