Skip to Content
Merck
All Photos(1)

Key Documents

SML0370

Sigma-Aldrich

Kartogenin

≥98% (HPLC)

Synonym(s):

2-[(Biphenyl-4-yl)carbamoyl]benzoic acid, 4′-Phenyl-phthalanilic acid (8CI), KGN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15NO3
CAS Number:
Molecular Weight:
317.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: >15 mg/mL

storage temp.

room temp

SMILES string

O=C(NC1=CC=C(C2=CC=CC=C2)C=C1)C(C=CC=C3)=C3C(O)=O

InChI

1S/C20H15NO3/c22-19(17-8-4-5-9-18(17)20(23)24)21-16-12-10-15(11-13-16)14-6-2-1-3-7-14/h1-13H,(H,21,22)(H,23,24)

InChI key

SLUINPGXGFUMLL-UHFFFAOYSA-N

Biochem/physiol Actions

Kartogenin induces the selective differentiation of multipotent mesenchymal stem cells (MSCs) into chondrocytes.
Kartogenin induces the selective differentiation of multipotent mesenchymal stem cells (MSCs) into chondrocytes. Kartogenin binds to filamin A, and disrupts the specific interaction between filamin A and CBFβ (core-binding factor β subunit). Apparently, kartogenin induces chondrogenesis by regulating the nuclear localization of CBFβ.
Kartogenin is a small heterocyclic compound. It is involved in regulating runt-related transcription factor 1 (RUNX1), which plays an important role in chondrocyte proliferation and survival.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yoshitaka Imamichi et al.
The Journal of clinical endocrinology and metabolism, 101(10), 3582-3591 (2016-07-19)
11-ketotestosterone (11-KT) is a novel class of active androgen. However, the detail of its synthesis remains unknown for humans. The objective of this study was to clarify the production and properties of 11-KT in human. Design, Participants, and Methods: Expression
Jun Zhao et al.
EBioMedicine, 36, 526-538 (2018-10-09)
Disordered folliculogenesis is a key feature of polycystic ovary syndrome (PCOS), but the underlying molecular mechanism remains unclear. Long non-coding RNA (lncRNA) expression in luteinized granulosa cells (hLGCs) derived from women with and without PCOS were analyzed using microarray and
Kartogenin induces cartilage-like tissue formation in tendon-bone junction
Zhang J and Wang J H C
Bone research, 2(1), 1-10 (2014)
Mei Zhang et al.
Cell death & disease, 8(10), e3088-e3088 (2017-10-06)
Oxidative stress impairs follicular development by inducing granulosa cell (GC) apoptosis, which involves enhancement of the transcriptional activity of the pro-apoptotic factor Forkhead box O1 (FoxO1). However, the mechanism by which oxidative stress promotes FoxO1 activity is still unclear. Here
Photo-cross-linked scaffold with kartogenin-encapsulated nanoparticles for cartilage regeneration
Shi D, et al.
ACS Nano, 10(1), 1292-1299 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service