Skip to Content
Merck
All Photos(1)

Key Documents

T7513

Sigma-Aldrich

Thrombin from bovine plasma

lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)

Synonym(s):

Factor IIa

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥2,000 NIH units/mg protein (E1%/280 = 19.5)

mol wt

heavy chain ~33 kDa
light chain ~5 kDa

UniProt accession no.

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Gene Information

cow ... F2(280685)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

Unit Definition

Activity is expressed in NIH units obtained by direct comparison to a NIH thrombin reference standard.

Physical form

Lyophilized from saline sodium citrate buffer, pH 6.5

Analysis Note

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard, Lot K.
The NIH assay procedure uses 0.2 mL of diluted plasma (1:1 with saline) as a substrate and 0.1 mL of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Other Notes

View more information on thrombin at www.sigma-aldrich.com/enzymeexplorer.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Stöhr et al.
Journal of molecular and cellular cardiology, 63, 189-198 (2013-07-31)
Myosin-binding protein C (Mybpc3)-targeted knock-in mice (KI) recapitulate typical aspects of human hypertrophic cardiomyopathy. We evaluated whether these functional alterations can be reproduced in engineered heart tissue (EHT) and yield novel mechanistic information on the function of cMyBP-C. EHTs were
Rosaria Ottanà et al.
Bioorganic chemistry, 92, 103211-103211 (2019-09-07)
Protein tyrosine phosphatase 1B (PTP1B) acts as a negative regulator of insulin and leptin signalling and is crucially involved in the development of type 2 diabetes mellitus, obesity, cancer and neurodegenerative diseases. Pursuing our efforts to identify PTP1B inhibitors endowed
Marco Castelli et al.
The Journal of biological chemistry, 279(43), 44731-44739 (2004-07-31)
Mitogen-activated protein (MAP) kinases play a central role in controlling a wide range of cellular functions following their activation by a variety of extracellular stimuli. MAP kinase phosphatases (MKPs) represent a subfamily of dual specificity phosphatases, which negatively regulate MAP
N J Royle et al.
Somatic cell and molecular genetics, 13(3), 285-292 (1987-05-01)
The gene for human prothrombin, or factor II (F2) has been assigned to 11p11-q12 by the combined use of a panel of somatic cell hybrid DNAs and in situ hybridization, using both cDNA and genomic probes. In addition, the cDNA
J Y Chueh et al.
AJNR. American journal of neuroradiology, 32(7), 1237-1244 (2011-05-21)
Mechanical behavior of the thromboembolus is one of the key factors that determine the efficacy of thrombectomy devices for revascularization in AIS. We characterized the mechanical properties and composition of thromboemboli from clinical cases and compared them with commonly used

Articles

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Thrombin Factor IIa is an endolytic serine protease that selectively cleaves the Arg--Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Protocols

Thrombin is an endolytic serine protease that selectively cleaves the Arg–Gly bonds of fibrinogen to form fibrin and release fibrinopeptides A and B.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service