Skip to Content
Merck
All Photos(1)

Key Documents

SML0353

Sigma-Aldrich

Azimilide dihydrochloride

≥97% (HPLC)

Synonym(s):

1-[[[5-(4-Chlorophenyl)-2-furanyl]methylene]amino]-3-[4-(4-methyl-1-piperazinyl)butyl]-2,4-imidazolidinedione dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28ClN5O3 · 2HCl
CAS Number:
Molecular Weight:
530.88
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Cl.Cl.CN1CCN(CCCCN2C(=O)CN(\N=C/c3ccc(o3)-c4cccc(Cl)c4)C2=O)CC1

InChI

1S/C23H28ClN5O3.2ClH/c1-26-11-13-27(14-12-26)9-2-3-10-28-22(30)17-29(23(28)31)25-16-20-7-8-21(32-20)18-5-4-6-19(24)15-18;;/h4-8,15-16H,2-3,9-14,17H2,1H3;2*1H/b25-16-;;

InChI key

SUHOTCCGXXEWJN-DSHYBBOZSA-N

Application

Azimilide dihydrochloride may be used in the electrophysiology experiments with human ether-a-go-go-related gene 1a (HERG1a) subunit.

Biochem/physiol Actions

Azimilide is a Class III antiarrhythmic; blocker of the delayed rectifier cardiac potassium channels; inhibits KV7.1 and KV11.1 potassium channels.
Azimilide is an inhibitor of human ether-a-go-go-related gene (HERG) channel. It displays a decrease in inhibitory effect in acidic pH conditions.
Azimilide is an investigational class III anti-arrhythmic drug that blocks fast and slow components of the delayed rectifier cardiac potassium channels. It inhibits KV7.1 and KV11.1 potassium channels. Azimilide′s block of K+ currents is relatively selective for IKr over IKs: It potently blocks the rapidly activating component of the delayed rectifier, IKr (IC50 0.4 mM), and inhibits IKs (IC50 3 mM) with nearly 10-fold less potency. At 10 mM, it does not block the inward rectifier K+ current. It blocks (10 mM) the L-type Ca2+ current (ICa) in a use-dependent manner.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donglin Guo et al.
Journal of cardiovascular electrophysiology, 18(2), 196-203 (2007-01-11)
The morphology of the mammalian cardiac action potential (AP) is an important factor in the susceptibility to drug-induced early afterdepolarizations (EADs) that may initiate torsade de pointes (TdP). AP triangulation has been shown to be an important predictor of drug-induced
S Sharma
The Journal of the Association of Physicians of India, 55 Suppl, 43-46 (2008-03-29)
The treatment of cardiac arrhythmias has undergone a sea change with the advent of catheter ablative procedures (radiofrequency ablation) and use of implantable cardioverter defibrillator (ICD). The antiarrhythmic drugs at times are used to prevent device related arrhythmia rather than
Atsushi Nishida et al.
Journal of pharmacological sciences, 105(3), 229-239 (2007-10-30)
Effects of azimilide, a class III antiarrhythmic drug, on the acetylcholine (ACh) receptor-operated K+ current (I K.ACh) and the delayed rectifier K+ current (IK) were examined in guinea-pig atrial cells using patch-clamp techniques. Effects of azimilide on experimental atrial fibrillation
Jarosław Kaźmierczak et al.
Pacing and clinical electrophysiology : PACE, 30(8), 1043-1046 (2007-08-03)
The presented case report describes a male patient with an implanted cardioverter-defibrillator (ICD) in whom a coadministration of ciprofloxacin and azimilide caused QT interval prolongation and multiple episodes of torsades de pointes (TdP) followed by ICD shocks (arrhythmic storm). The
Philippe Chevalier et al.
Journal of cardiovascular pharmacology, 50(6), 629-632 (2007-12-20)
The effects of chronic oral azimilide therapy on the ventricular defibrillation threshold (DFT) during ischemia are unknown. The effects of azimilide on defibrillation efficacy under ischemic condition were investigated in a closed-chest animal model. Azimilide (20 mg/kg/d) was administered orally

Articles

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service