Skip to Content
Merck
All Photos(1)

Key Documents

SML0346

Sigma-Aldrich

Alosetron hydrochloride

≥98% (HPLC)

Synonym(s):

2,3,4,5-Tetrahydro-5-methyl-2-[(5-methyl-1H-imidazol-4-yl)methyl]-1H-pyrido[4,3-b]indol-1-one hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N4O·HCl
CAS Number:
Molecular Weight:
330.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: ≥5 mg/mL at warmed

storage temp.

2-8°C

SMILES string

CN1C2=C(C=CC=C2)C3=C1CCN(CC4=C(C)N=CN4)C3=O.Cl

InChI

1S/C17H18N4O.ClH/c1-11-13(19-10-18-11)9-21-8-7-15-16(17(21)22)12-5-3-4-6-14(12)20(15)2;/h3-6,10H,7-9H2,1-2H3,(H,18,19);1H

InChI key

FNYQZOVOVDSGJH-UHFFFAOYSA-N

Gene Information

human ... HTR3A(3359)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Alosetron is a potent and highly selective antagonist of serotonin 5-HT3 receptors, nonselective cation channels found predominantly in the enteric nervous system of the gastrointestinal tract. These receptors are involved in the regulation of visceral pain, colonic transit and GI secretions that can contribute to the pathophysiology of irritable bowel syndrome (IBS). Alosetron is used clinically for treatment of women with severe diarrhea-predominant IBS.
Alosetron is a potent and highly selective antagonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James H Lewis
Drug safety, 34(7), 545-565 (2011-06-15)
Ischaemic colitis (IC) is the most common form of ischaemic injury to the gastrointestinal (GI) tract. IC typically presents with the sudden onset of lower abdominal pain, cramping and rectal bleeding, and is usually self-limited with low morbidity, although it
Lin Chang et al.
The American journal of gastroenterology, 105(4), 866-875 (2010-03-04)
Alosetron is a potent, selective 5-HT(3) receptor antagonist prescribed for women with severe diarrhea-predominant irritable bowel syndrome (IBS-D) under a risk management plan (RMP). The RMP was implemented following cases of ischemic colitis (IC) and complications of constipation (CoC) associated
Andrea Fantuzzi et al.
Analytical chemistry, 82(24), 10222-10227 (2010-11-26)
This paper is the first report of a P450-electrode in a microfluidic format. A 30 μL microfluidic cell was made in poly(methyl methacrylate) containing the inlet, outlet, and reaction chamber with two electrode strips, one of which contains the human
Eric Shah et al.
The American journal of medicine, 125(4), 381-393 (2012-03-27)
Current treatment options for irritable bowel syndrome are limited and often poorly studied. A select few drugs have been studied in irritable bowel syndrome, and the number needed to treat is frequently used to assess the relative efficacy of these
Larry D Lynd et al.
Value in health : the journal of the International Society for Pharmacoeconomics and Outcomes Research, 13(4), 411-417 (2009-09-12)
There is consensus that a more transparent, explicit, and rigorous approach to benefit-risk evaluation is required. The objective of this study is to evaluate the incremental net benefit (INB) framework for undertaking quantitative benefit-risk assessment by performing a quantitative benefit-risk

Articles

Human epithelial intestinal colonic organoids can be used as an alternative to Caco-2 drug permeability assays for drug screening and compound toxicity testing.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

We offer many products related to serotonin receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service