Skip to Content
Merck
All Photos(1)

Key Documents

SML0319

Sigma-Aldrich

Caramiphen hydrochloride

≥98% (HPLC)

Synonym(s):

2-(Diethylamino)ethyl 1-phenylcyclopentane-1-carboxylate hydrochloride, Caramiphenium chloride salt, G 2747, Panparnit, Parpanil, Parpanit, Pentaphen, Pentaphene hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H27NO2 · HCl
CAS Number:
Molecular Weight:
325.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CCN(CC)CCOC(=O)C1(CCCC1)c2ccccc2

InChI

1S/C18H27NO2.ClH/c1-3-19(4-2)14-15-21-17(20)18(12-8-9-13-18)16-10-6-5-7-11-16;/h5-7,10-11H,3-4,8-9,12-15H2,1-2H3;1H

InChI key

MUPNXGNOIBYHSG-UHFFFAOYSA-N

Application

Caramiphen hydrochloride has been used to test its inhibitory effect on voltage-gated Na+ currents using the patch-clamp method.

Biochem/physiol Actions

Caramiphen hydrochloride is non-narcotic and possesses many biological functionalities of being antitussive, anticonvulsive, and neuroprotective. It induces sensory block and displays less toxicity when compared to bupivacaine. The local anesthetic functionality of caramiphen is mediated by its inhibitory effect on voltage-gated Na+ currents.
Caramiphen is an anti-muscarinic similar to atropine; muscarinic acetylcholine receptor antagonist. Caramiphen is commonly used as a cough suppresant, but is also administered for the treatment of Parkinson′s disease. The compound has also been shown to have anti-depressant properties that may involve inhibition of G protein-coupled inwardly rectifying potassium channel GIRK.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M R Domenici et al.
General pharmacology, 24(4), 991-998 (1993-07-01)
1. The effects of the non-opioid antitussives caramiphen and carbetapentane and of the anticonvulsants 5,5-diphenylhydantoin and MK 801 were tested towards hypoxia-induced electrical changes in rat hippocampal slices. 2. The incidence of appearance of hypoxia-induced epileptiform bursting was significantly decreased
G Diana et al.
Life sciences, 52(19), 1547-1557 (1993-01-01)
The effects of the non-opioid oral antitussives dextromethorphan (DM) and caramiphen (CP) were tested against the behavioural and EEG effects elicited by the N-methyl-D-aspartate (NMDA) antagonists dizocilpine (MK 801) and phencyclidine (PCP) in rats and mice. PCP (1.25-10 mg/kg i.p.)
Yuk-Man Leung et al.
European journal of pharmacology, 746, 213-220 (2014-12-03)
The underlying mechanisms for the action of caramiphen used in local anesthesia are not well understood. The purpose of this study was to evaluate the block of caramiphen on voltage-gated Na⁺ channels and in spinal anesthesia. We investigated the effect
Trond Myhrer et al.
Neurotoxicology, 34, 128-134 (2012-11-21)
Microinfusion of anticonvulsants into the perirhinal cortex through 1 guide cannula in each hemisphere only invades a small area of this seizure controlling site in rats exposed to soman. The purpose of the present study was to examine whether infusions
M Gavend et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 49(10), 456-464 (1995-01-01)
Male Sprague-Dawley rats were trained to discriminate dextromethorphan (DM, 30 mg/kg, ip) from saline using a standard two-lever, fixed ratio 10, food reinforcement procedure. The DM-saline discrimination was acquired, and a range of doses of DM produced a dose-related generalization

Articles

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

Muscarinic acetylcholine receptors mediate acetylcholine actions in CNS and non-nervous tissues, crucial for cell signaling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service