Skip to Content
Merck
All Photos(1)

Documents

S7882

Sigma-Aldrich

(−)-Scopolamine N-butyl bromide

≥98% (TLC), powder

Synonym(s):

(−)-N-Butylscopolamine bromide, Hyoscine N-butyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30BrNO4
CAS Number:
Molecular Weight:
440.37
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

optical activity

[α]25/D −20.8°, c = 3 in H2O(lit.)

color

white

solubility

H2O: 50 mg/mL

SMILES string

[Br-].[H][C@@]12C[C@H](C[C@@]([H])([C@]3([H])O[C@]13[H])[N+]2(C)CCCC)OC(=O)[C@H](CO)c4ccccc4

InChI

1S/C21H30NO4.BrH/c1-3-4-10-22(2)17-11-15(12-18(22)20-19(17)26-20)25-21(24)16(13-23)14-8-6-5-7-9-14;/h5-9,15-20,23H,3-4,10-13H2,1-2H3;1H/q+1;/p-1/t15-,16-,17-,18+,19-,20+,22?;/m1./s1

InChI key

HOZOZZFCZRXYEK-GSWUYBTGSA-M

Looking for similar products? Visit Product Comparison Guide

Application

(−)-Scopolamine N-butyl bromide was used as standard in designing a procedure for quantification of compounds using CE-MS.8

Biochem/physiol Actions

Competitive muscarinic acetylcholine receptor antagonist; antispasmodic.
Scopolamine is an anti-muscarinic and cholinolytic alkaloid that inhibits parasympathetic-cholinergic system.5 The central effects include hallucinations, disorientation, restlessness and euphoria.6 Scopolamine disturb the stimulus detection performance in rats by interfering with the reticular cholinergic pathways.7

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jens Ohnesorge et al.
Electrophoresis, 26(12), 2360-2375 (2005-06-01)
Different approaches were chosen to examine ionization reproducibility of analytes after separation by capillary electrophoresis-mass spectrometry (CE-MS) in a commercially available sheath-flow electrospray interface. For this task three different standard samples were examined. Sample 1 contained neostigmine bromide (cationic), paracetamol
S Lin et al.
Digestive diseases and sciences, 42(5), 907-914 (1997-05-01)
Expression of muscarinic receptor subtypes in rat gastric smooth muscle was examined with reverse transcriptase-polymerase chain reaction (RT-PCR). Under the condition for detecting the messages of m1-m4 subtypes in brain, atrium, and gastric mucosa, only the fragments of m2 and
K Kurahashi et al.
Life sciences, 61(8), 831-838 (1997-01-01)
The inhibitory effects of atropine, cimetropium, pirenzepine and N-butylscopolamine on the vagal afferent gastric excitatory response in cats under anesthesia with pentobarbital sodium and infusion of gallamine were examined. Electrical stimulation of vagal trunk in left side (10 Hz in
A J Morton et al.
Equine veterinary journal. Supplement, (39)(39), 117-122 (2011-08-04)
N-butylscopolammonium bromide (NBB) and xylazine are commonly used medications for the treatment of spasmodic colic and other forms of abdominal pain in horses. Both NBB and xylazine exert significant effects on the cardiovascular system and other vital systems of horses.
J Künzel et al.
Reproductive biomedicine online, 23(1), 132-140 (2011-07-16)
To compare the effects of neurotropic and musculotropic spasmolytic drugs in isolated swine uterus specimens, 80 swine uteri were perfused using an established model for preserving a viable organ that responds to oxytocic hormones and spasmolytic drugs. An intrauterine catheter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service