Skip to Content
Merck
All Photos(1)

Key Documents

SML0310

Sigma-Aldrich

Tryptanthrin

≥98% (HPLC)

Synonym(s):

Couroupitine A, Indolo(2,1-b)quinazoline-6,12-dione, NSC 349447, Tryptanthrine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8N2O2
CAS Number:
Molecular Weight:
248.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

faintly yellow to dark yellow

solubility

DMSO: 2 mg/mL (warmed)

storage temp.

2-8°C

SMILES string

O=C1C2=NC3=CC=CC=C3C(N2C4=C1C=CC=C4)=O

InChI

1S/C15H8N2O2/c18-13-10-6-2-4-8-12(10)17-14(13)16-11-7-3-1-5-9(11)15(17)19/h1-8H

InChI key

VQQVWGVXDIPORV-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

Tryptanthrin is a plant alkaloid with anti-inflammatory and anti cancer activities.
Tryptanthrin is a plant alkaloid with anti-inflammatory and anti cancer activities. Tryptanthrin blocks leukotriene production in neutrophils and in whole blood assays, and in an in vivo rat pleurisy model. The compound also inhibits P-glycoprotein, and sensitizes resistant cancer cell lines to killing by cytotoxic agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jae-Min Hwang et al.
Journal of natural products, 76(3), 354-367 (2013-01-31)
The natural product tryptanthrin (1a) represents a potential lead for new tuberculosis (TB) drugs since tryptanthrin and its synthetic analogues possess potent in vitro activity against Mycobacterium tuberculosis (Mtb). However, in spite of their in vitro activity, none of these
C Vlachos et al.
The British journal of dermatology, 167(3), 496-505 (2012-04-27)
The aryl hydrocarbon receptor (AhR) is a nuclear receptor and transcriptional regulator with pleiotropic effects. The production of potent AhR ligands by Malassezia yeasts, such as indirubin, indolo[3,2-b]carbazole (ICZ), tryptanthrin and malassezin, has been associated with the pathogenesis of seborrhoeic
Man Hagiyama et al.
Experimental and therapeutic medicine, 23(4), 274-274 (2022-03-08)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) uses its S1 spike protein to bind to angiotensin-converting enzyme 2 (ACE2) on human cells in the first step of cell entry. Tryptanthrin, extracted from leaves of the indigo plant, Polygonum tinctorium, using
Bing-Chung Liau et al.
Journal of pharmaceutical and biomedical analysis, 43(1), 346-351 (2006-08-04)
A rapid, selective, and sensitive LC-APCI-MS method is developed in this study for detecting and analyzing tryptanthrin, indigo, and indirubin in daqingye and banlangen, which are, respectively, the leaves and roots of Isatis indigotica and Strobilanthes cusia in traditional Chinese
Hui-Man Cheng et al.
BMC complementary and alternative medicine, 17(1), 439-439 (2017-09-04)
Indigo naturalis is a Traditional Chinese Medicine (TCM) ingredient long-recognized as a therapy for several inflammatory conditions, including psoriasis. However, its mechanism is unknown due to lack of knowledge about the responsible chemical entity. We took a different approach to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service