Skip to Content
Merck
All Photos(1)

Key Documents

229296

Sigma-Aldrich

Indigo

synthetic, Dye content 95 %

Synonym(s):

Indigo blue, Indigotin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10N2O2
CAS Number:
Molecular Weight:
262.26
Colour Index Number:
73000
Beilstein:
88275
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 95%

technique(s)

titration: suitable

mp

>300 °C (lit.)

density

1.35 g/cm3 at 20 °C

λmax

602 nm

ε (extinction coefficient)

≥8000 at 599-605 nm in chloroform

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O=C1C(\Nc2ccccc12)=C3/Nc4ccccc4C3=O

InChI

1S/C16H10N2O2/c19-15-9-5-1-3-7-11(9)17-13(15)14-16(20)10-6-2-4-8-12(10)18-14/h1-8,17-18H/b14-13+

InChI key

COHYTHOBJLSHDF-BUHFOSPRSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Application

Indigo has been used as a dye to measure food consumption in Drosophila. It has also been used to investigate the ability of fungal strains in the degradation and decolorization of indigo dye.

Biochem/physiol Actions

Indigo is a rich blue color dye. It can be extracted from plants of genus Indigofera and can also be produced synthetically. Indigo is a vat textile dye, widely used on cellulosic fibers like cotton. Additionally, it is also used to inhibit corrosion of mild steel in aerated sulfuric acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of fungal decolorization of azo and anthraquinone dyes via Box-Behnken design
Papadopoulou K , et al.
International Biodeterioration & Biodegradation, 77, 31-38 (2013)
Indigo: Cultivate, dye, create (2018)
Insulin-producing cells in the Drosophila brain also express satiety-inducing cholecystokinin-like peptide, drosulfakinin
Soderberg JAE, et al.
Frontiers in Endocrinology, 3, 109-109 (2012)
Decolorization of textile indigo dye by ligninolytic fungi
Balan DSL and Monteiro RTR
Journal of Biotechnology, 89(2-3), 141-145 (2001)
Inhibition of mild steel corrosion in sulphuric acid using indigo dye and synergistic halide additives
Oguzie EE, et al.
Materials Chemistry and Physics, 84, 363-368 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service