Skip to Content
Merck
All Photos(1)

Key Documents

SML0021

Sigma-Aldrich

PDI inhibitor 16F16

≥98% (HPLC)

Synonym(s):

2-(2-Chloroacetyl)-2,3,4,9-tetrahydro-1-methyl-1H-pyrido[3,4-b]indole-1-carboxylic acid methyl ester, Methyl 2-(2-chloroacetyl)-1-methyl-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17ClN2O3
CAS Number:
Molecular Weight:
320.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: 12 mg/mL, clear

storage temp.

2-8°C

SMILES string

COC(=O)C1(C)N(CCc2c1[nH]c3ccccc23)C(=O)CCl

InChI

1S/C16H17ClN2O3/c1-16(15(21)22-2)14-11(7-8-19(16)13(20)9-17)10-5-3-4-6-12(10)18-14/h3-6,18H,7-9H2,1-2H3

InChI key

BCSIRYFYAKLJDK-UHFFFAOYSA-N

Application

PDI inhibitor 16F16 has been used to inhibit PDI (protein disulfide isomerase) function to examine the functional conservation of PDIs in human embryonic kidney cells.
PDI inhibitor 16F16 may be used in protein disulfide isomerase-mediated cell signaling studies.

Biochem/physiol Actions

16F16 is a protein disulfide isomerase (PDI) inhibitor, first identified in a screen for compounds that prevent apoptosis induced by mutant huntingtin protein. 16F16 not only suppressed apoptosis induced by the misfolded protein mutant hungtingtin, it also protected rat neurons from cell death triggered by Aβ peptide. The actions of this inhibitor helped to identify a new mechanism in which a cell death pathway is regulated by protein misfolding via PDI upregulation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Protein Disulfide Isomerase Controls Neuronal Migration through Regulation of Wnt Secretion
Torpe N, et al.
Cell Reports, 26(12), 3183-3190 (2019)
Nanna Torpe et al.
Cell reports, 26(12), 3183-3190 (2019-03-21)
Appropriate Wnt morphogen secretion is required to control animal development and homeostasis. Although correct Wnt globular structure is essential for secretion, proteins that directly mediate Wnt folding and maturation remain uncharacterized. Here, we report that protein disulfide isomerase-1 (PDI-1), a protein-folding
Ying Yuan et al.
Materials today. Bio, 21, 100697-100697 (2023-06-22)
Anastomotic thrombosis prevalently causes anastomosis failure, accompanied with ischemia and necrosis, the early diagnosis of which is restricted by inherent shortcomings of traditional imaging techniques in clinic and lack of appropriate prodromal biomarkers for thrombosis initiation. Herein, a fresh thrombus-specific
Nayeli Aguilar-Hernández et al.
Viruses, 13(1) (2021-01-06)
Although human astroviruses (HAstVs) are important agents of gastroenteritis in young children, the studies aimed at characterizing their biology have been limited, in particular regarding their cell entry process. It has been shown that HAstV serotype 8 enters human cells
Yuanyuan Yan et al.
Cell death and differentiation, 29(9), 1769-1789 (2022-03-19)
Hypoxic tumor microenvironment (TME) plays critical roles in induction of cancer stem cell-like phenotype in breast cancer and contribute to chemoresistance. However, the mechanism underlying stemness reprogramming of breast cancer cells (BCs) by hypoxic TME remains largely unknown. In the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service