Skip to Content
Merck
All Photos(1)

Key Documents

D95654

Sigma-Aldrich

Diethyl fumarate

98%

Synonym(s):

(2E)-2-Butenedioic acid diethyl ester, (E)-But-2-enedioic acid diethyl ester, Diethyl (E)-but-2-enedioate, Diethyl fumaric acid ester, Ethyl fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCH=CHCOOC2H5
CAS Number:
Molecular Weight:
172.18
Beilstein:
775347
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.44 (lit.)

bp

218-219 °C (lit.)

mp

1-2 °C (lit.)

density

1.052 g/mL at 25 °C (lit.)

SMILES string

[H]\C(=C(\[H])C(=O)OCC)C(=O)OCC

InChI

1S/C8H12O4/c1-3-11-7(9)5-6-8(10)12-4-2/h5-6H,3-4H2,1-2H3/b6-5+

InChI key

IEPRKVQEAMIZSS-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl fumarate is an electrophilic compound, commonly used in the Diels-Alder reaction.

Application


  • Relaxation and Amorphous Structure of Polymers: This study examines the behavior of polymers with rigid fumarate segments, including diethyl fumarate, and their impact on the relaxation and amorphous structures, enhancing polymer design and processing (Suzuki et al., 2022).

  • Gradient versus End-Capped Degradable Polymer Sequence Variations: The research explores the mechanical properties of photochemically 3D-printed substrates using diethyl fumarate-based polymers, presenting new possibilities in creating tailored biomaterials for medical applications (Shin and Becker, 2022).

  • Novel Curcumin-Diethyl Fumarate Hybrid for Parkinson′s Disease: This study investigates a hybrid molecule combining curcumin and diethyl fumarate, highlighting its dual function as a GSK-3β inhibitor and Nrf2 inducer, which could be significant for Parkinson′s disease treatment strategies (Di Martino et al., 2020).

  • Synthesis and 3D Printing of PEG-Poly(propylene fumarate) Copolymers: This article describes the synthesis and application of diethyl fumarate in the creation of novel diblock and triblock copolymer hydrogels for 3D printing, expanding the utility in biomedicine and tissue engineering (Dilla et al., 2018).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Recent advances in understanding NRF2 as a druggable target: development of pro-electrophilic and non-covalent NRF2 activators to overcome systemic side effects of electrophilic drugs like dimethyl fumarate
Takumi S et al.
F1000Research, 6 (2017)
B Sebök et al.
Skin pharmacology : the official journal of the Skin Pharmacology Society, 9(2), 99-103 (1996-01-01)
Fumaric acid, fumaric acid dimethylester, and the dithranol derivative C4-lactone were studied in the mouse tail test to evaluate their effects on epidermal cell differentiation compared with other topical antipsoriatic drugs, such as betamethasone, calcipotriol, and dithranol. Mouse tails were
Jin Woo Lee et al.
Biomaterials, 32(3), 744-752 (2010-10-12)
Bony defects have been three-dimensionally (3D) created in many clinical circumstances; however, many defects cannot be reconstructed because most of the current bony substitutes cannot provide the necessary exact 3D structure. Therefore, to overcome this limitation, a 3D scaffold with
A Lahti et al.
Contact dermatitis, 16(3), 133-135 (1987-03-01)
To investigate the mechanisms of non-immunologic contact urticaria (NICU), the effects of 1g + 1g of acetylsalicylic acid (ASA) on contact reactions to methyl nicotinate, diethyl fumarate, benzoic acid, cinnamic acid, cinnamic aldehyde and dimethyl sulfoxide were studied in 21
Pattama Saisaha et al.
Organic & biomolecular chemistry, 8(19), 4444-4450 (2010-08-18)
A practical method for the multigram scale selective cis-dihydroxylation of electron deficient alkenes such as diethyl fumarate and N-alkyl and N-aryl-maleimides using H(2)O(2) is described. High turnovers (>1000) can be achieved with this efficient manganese based catalyst system, prepared in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service