Skip to Content
Merck
All Photos(1)

Key Documents

SML0011

Sigma-Aldrich

VAHA

≥98% (HPLC)

Synonym(s):

VAHA, Valproyl hydroxamic acid, VPA-HA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17NO2
CAS Number:
Molecular Weight:
159.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: ≥25 mg/mL

storage temp.

2-8°C

SMILES string

CCCC(CCC)C(=O)NO

InChI

1S/C8H17NO2/c1-3-5-7(6-4-2)8(10)9-11/h7,11H,3-6H2,1-2H3,(H,9,10)

InChI key

ROJGIRXXBBBMPL-UHFFFAOYSA-N

Application

VAHA may be used in cell signaling studies.

Biochem/physiol Actions

Hydroxamic acid derivatives of valproic acid exhibit anticonvulsant activity with no teratogenic activity in mouse neural tube defect model. It is effective in the treatment of bipolar disorders.
VAHA (Valproyl hydroxamic acid) is an HDAC inhibitor with less activity than valproic acid against Class I enzymes but much greater Class II activity

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Libert et al.
Biomedical & environmental mass spectrometry, 13(11), 599-603 (1986-11-01)
A previously unreported substance, detected by gas chromatography after oximation of urine from patients receiving valproic acid, was isolated and analysed by mass spectrometry and nuclear magnetic resonance spectroscopy. It was identified as the hydroxamate of valproic acid.
B J Eickholt et al.
Molecular pharmacology, 67(5), 1426-1433 (2005-02-03)
Inositol-1,4,5-trisphosphate (InsP3) depletion has been implicated in the therapeutic action of bipolar disorder drugs, including valproic acid (VPA). It is not currently known whether the effect of VPA on InsP3 depletion is related to the deleterious effects of teratogenicity or
Ute Gravemann et al.
Neurotoxicology and teratology, 30(5), 390-394 (2008-05-06)
Fluorinated and non-fluorinated valproic acid (VPA) analogues with hydroxamic acid moieties were tested for their teratogenic, anticonvulsant and neurotoxic potencies in mice. Compounds were synthesized from their corresponding acids. The induction of neural tube defects (exencephaly) of the resulting hydroxamates

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service