Skip to Content
Merck
All Photos(1)

Key Documents

559418

Sigma-Aldrich

SBHA

A cell-permeable bishydroxamic acid histone deacetylase (HDAC) inhibitor with anti-tumor properties.

Synonym(s):

SBHA, Suberic Bishydroxamate, Suberoyl Bishydroxamic Acid, HDAC Inhibitor XII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O4
CAS Number:
Molecular Weight:
204.22
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(O)C(=O)CCCCCCC(=O)NO

InChI

1S/C8H16N2O4/c11-7(9-13)5-3-1-2-4-6-8(12)10-14/h13-14H,1-6H2,(H,9,11)(H,10,12)

InChI key

IDQPVOFTURLJPT-UHFFFAOYSA-N

General description

A cell-permeable bishydroxamic acid histone deacetylase (HDAC) inhibitor with anti-tumor properties. Reported to cause an increase in acetylated histone H4 in MEL cells. Also reported to inhibit human HDAC1 and HDAC3 activities with a similar potency (ID50 ~ 250-300 nM). Belongs to the hybrid polar class of compounds that induce both differentiation of transformed cells and apoptosis. Shown to suppress the growth of proliferating keratinocytes and squamous cell carcinoma cells (IC50 = 11.57 µM and 5.39 µM, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
human HDAC1 and HDAC3 activities
Product does not compete with ATP.
Reversible: no
Target IC50: 11.57 µM and 5.39 µM in suppressing the growth of proliferating keratinocytes and squamous cell carcinoma cells, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.





From Catalog:

Desc. Field- added "cell-permeable"

Other Notes

Brinkmann, H., et al. 2001. J. Biol. Chem.276, 22491.
Richon, V.M., et al. 1998. Proc. Natl. Acad. Sci. USA 95, 3003.
Richon, V.M., et al. 1996. Proc. Natl. Acad. Sci. USA93, 5705.
Marks, P.A., 1994. Proc. Natl. Acad. Sci. USA91, 10251.

Legal Information

Not available for sale in the United States.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service