Skip to Content
Merck
All Photos(4)

Key Documents

SHH0001

Sigma-Aldrich

Novec 7000 Engineered Fluid

greener alternative

≥99.5% purity, liquid

Synonym(s):

1-Methoxyheptafluoropropane, Methyl perfluoropropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2CF2OCH3
CAS Number:
Molecular Weight:
200.05
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Novec 7000 Engineered Fluid, ≥99.5%

Assay

≥99.5%

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

titration: suitable

color

colorless to yellow

mp

-122.49 °C

density

1.40 g/mL at 25 °C

application(s)

diagnostic assay manufacturing
hematology
histology

greener alternative category

storage temp.

room temp

SMILES string

FC(C(F)(F)OC)(F)C(F)(F)F

InChI

1S/C4H3F7O/c1-12-4(10,11)2(5,6)3(7,8)9/h1H3

InChI key

NOPJRYAFUXTDLX-UHFFFAOYSA-N

Application

  • Low Global Warming Potential/GWP (530, 100-year ITH)
  • Excellent dielectric properties: In event of leakage or other failure, will not damage electronic equipment
  • Zero ozone depletion potential (ODP)
  • Good materials compatibility
  • Low toxicity
  • Non-flammable
  • Non-corrosive
  • Good thermal stability
  • Useful at extreme low temperatures
  • Viscosity is less than 20 cSt at -120 °C
Cryofreezing solution

Packaging

Packaged and distributed by Sigma-Aldrich

Legal Information

Novec is a trademark of 3M Company

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service