287571
Bis(2,2,2-trifluoroethyl) ether
98%
Synonym(s):
2,2,2-Trifluoroethyl ether, Flurothyl
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CF3CH2)2O
CAS Number:
Molecular Weight:
182.06
Beilstein:
1811927
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.300 (lit.)
bp
62-63 °C (lit.)
density
1.404 g/mL at 25 °C (lit.)
functional group
ether
fluoro
SMILES string
FC(F)(F)COCC(F)(F)F
InChI
1S/C4H4F6O/c5-3(6,7)1-11-2-4(8,9)10/h1-2H2
InChI key
KGPPDNUWZNWPSI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Bis(2,2,2-trifluoroethyl) ether was used to diminish self-discharge of Li-S cells having both low- and high-sulfur-loading sulfur cathodes.
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
33.8 °F - closed cup
Flash Point(C)
1 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Stacey B B Dutton et al.
Epilepsia, 52(11), 2050-2056 (2011-08-02)
We evaluated the ability of the ketogenic diet (KD) to improve thresholds to flurothyl-induced seizures in two mouse lines with Scn1a mutations: one that models Dravet syndrome (DS) and another that models genetic (generalized) epilepsy with febrile seizures plus (GEFS+).
Yogendrasinh H Raol et al.
Annals of neurology, 65(3), 326-336 (2009-04-01)
Neonatal seizures occur frequently, are often refractory to anticonvulsants, and are associated with considerable morbidity and mortality. Genetic and electrophysiological evidence indicates that KCNQ voltage-gated potassium channels are critical regulators of neonatal brain excitability. This study tests the hypothesis that
Li-Li Li et al.
Frontiers in neurology, 9, 802-802 (2018-10-16)
The role of leptin in the pathogenesis of epilepsy is getting more and more attention in clinical and basic research. Although there are data indicating neuroprotective effects of elevated serum/brain leptin levels following acute seizures, no study to date has
David N Ruskin et al.
PloS one, 8(6), e65021-e65021 (2013-06-12)
Autism spectrum disorders share three core symptoms: impaired sociability, repetitive behaviors and communication deficits. Incidence is rising, and current treatments are inadequate. Seizures are a common comorbidity, and since the 1920's a high-fat, low-carbohydrate ketogenic diet has been used to
Marcella M Lucas et al.
Brain : a journal of neurology, 134(Pt 6), 1684-1693 (2011-05-24)
One of the most common and serious co-morbidities in patients with epilepsy is cognitive impairment. While early-life seizures are considered a major cause for cognitive impairment, it is not known whether it is the seizures, the underlying neurological substrate or
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service