Skip to Content
Merck
All Photos(1)

Key Documents

SHC216

Sigma-Aldrich

MISSION® TRC2 pLKO.5-puro Non-Target shRNA Control Plasmid DNA

Targets no known genes from any species

Synonym(s):

MISSION® Control Vectors, negative control, negative shRNA control, non-target control, non-target shRNA, non-target shRNA control, shRNA control

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106609
NACRES:
NA.51

Quality Level

product line

MISSION®

concentration

500 ng/μL in TE buffer; DNA (10μg of plasmid DNA)

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The MISSION® TRC2 pLKO.5-puro Non-Target shRNA Control Plasmid DNA is a lentivirus plasmid vector. The vector contains an shRNA insert that does not target any known genes from any species, making it useful as a negative control in experiments using the MISSION® shRNA library clones. This allows one to examine the effect of transfection of a short-hairpin on gene expression and interpret the knockdown effect seen with shRNA clones. Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells respectively. In addition, self-inactivating replication incompetent viral particles can be produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids. The Non-Target shRNA Control Plasmid DNA is provided as 10 μg of plasmid DNA in Tris-EDTA (TE) buffer at a concentration of 500 ng/μl.

Application

To see more application data, protocols, vector maps visit sigma.com/shrna.

Legal Information

Use of this product is subject to one or more license agreements. For details, please see http://sigmaaldrich.com/missionlicense.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Pu et al.
The Journal of cell biology, 216(12), 4183-4197 (2017-10-11)
Lysosomes play key roles in the cellular response to amino acid availability. Depletion of amino acids from the medium turns off a signaling pathway involving the Ragulator complex and the Rag guanosine triphosphatases (GTPases), causing release of the inactive mammalian
Deanna M Santer et al.
Journal of immunological methods, 445, 15-22 (2017-03-10)
Type III interferons (IFN-lambdas) are important antiviral cytokines that also modulate immune responses acting through a unique IFN-λR1/IL-10R2 heterodimeric receptor. Conflicting data has been reported for which cells express the IFN-λR1 subunit and directly respond to IFN-λs. In this study
Cancer therapies activate RIG-I-like receptor pathway through endogenous non-coding RNAs.
Ranoa DR
Oncotarget, 7(18) (2016)
Simon J Conn et al.
Cell, 160(6), 1125-1134 (2015-03-15)
Circular RNAs (circRNAs), formed by non-sequential back-splicing of pre-mRNA transcripts, are a widespread form of non-coding RNA in animal cells. However, it is unclear whether the majority of circRNAs represent splicing by-products without function or are produced in a regulated
Adam L Green et al.
Oncogene, 39(11), 2305-2327 (2019-12-18)
High-grade gliomas (HGG) afflict both children and adults and respond poorly to current therapies. Epigenetic regulators have a role in gliomagenesis, but a broad, functional investigation of the impact and role of specific epigenetic targets has not been undertaken. Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service