Skip to Content
Merck
All Photos(1)

Key Documents

SHP001

Sigma-Aldrich

MISSION® Lentiviral Packaging Mix

Production of replication incompetent viral particles

Synonym(s):

Lentiviral Packaging, Lentiviral Packaging Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

product line

MISSION®

storage temp.

−20°C

General description

The MISSION® shRNA product line is centered on a transfer vector-based RNAi library against annotated mouse and human genes that can be used in conjunction with this Lentiviral Packaging Mix. In this packaging system, the pseudo-typing with VSV-G broadens the viral tropism associated with the virus. Therefore, these lentiviral particles can efficiently deliver the transfer sequence of interest to a wide variety of cell types, including primary and non-dividing cells.

The MISSION® Lentiviral Packaging Mix is an optimized formulation of two plasmids expressing the key HIV packaging genes and a heterologous viral envelope gene.

Lentiviral particles are generated from three main components:
  • The packaging vector, which contains the minimal set of lentiviral genes required to generate the virion structural proteins and packaging functions.
  • The vesicular stomatitis virus G-protein envelope vector, which provides the heterologous envelope for pseudotyping.
  • The shRNA transfer vector, which contains the sequence of interest as well as the cis acting sequences necessary for RNA production and packaging.

The Lentiviral Packaging Mix contains the first two components; it is designed to be co-transfected along with a compatible lentiviral transfer vector in order to create high-titer pseudo-typed lentiviral particles used for downstream transduction applications.

Application

MISSION® Lentiviral Packaging Mix has been used to incubate plasmids before addition to human embryonic kidney cells 293 expressing the large T antigen (293T cells) and to pack lentivirus into 293T cells.

Other Notes

Lentiviral particles are produced using a third-generation packaging system, which has many features that lead to enhanced biosafety. Recombinant lentiviruses produced with the MISSION Lentiviral Packaging Mix have not been shown to produce replication competent viral particles because of designed safety features. Users should consult and observe their own institutional guidelines before working with such viral systems.

Legal Information

Use of this product is subject to one or more license agreements. For details, please see http://sigmaaldrich.com/missionlicense .
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenwen Chien et al.
Journal of Cancer, 9(24), 4762-4773 (2018-12-28)
This study is an unbiased genomic screen to obtain functional targets for increased effectiveness of dasatinib in pancreatic cancer. Dasatinib, a multi-targeted tyrosine kinase inhibitor, is used in clinical trials for treatment of pancreatic cancer; however, intrinsic and acquired resistance
NIH Guidelines for Research Involving Recombinant DNA Molecules
N.I.H. Memorandum (2002)
Deniz Simsek et al.
PLoS genetics, 7(6), e1002080-e1002080 (2011-06-10)
Nonhomologous end-joining (NHEJ) is the primary DNA repair pathway thought to underlie chromosomal translocations and other genomic rearrangements in somatic cells. The canonical NHEJ pathway, including DNA ligase IV (Lig4), suppresses genomic instability and chromosomal translocations, leading to the notion
Heterogenous ribonucleoprotein A18 (hnRNP A18) promotes tumor growth by increasing protein translation of selected transcripts in cancer cells.
Chang ET
Oncotarget, 7(9), 10578-10593 (2016)
Helen He et al.
Journal of biomarkers, 2016, 1274603-1274603 (2016-06-02)
Objective. Use of tyramide signal amplification (TSA) to detect autophagy biomarkers in formalin fixed and paraffin embedded (FFPE) xenograft tissue. Materials and Methods. Autophagy marker regulation was studied in xenograft tissues using Amp HQ IHC and standard IHC methods. Results.

Articles

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

MISSION shRNA reduce the expression of specific target genes by targeting the specific mRNA therefore reducing the corresponding protein expression.

See All

Protocols

Preparation of the Lentiviral Transduction Particles Using Packaging Plasmid Mix

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service