Skip to Content
Merck
All Photos(3)

Documents

SAE0090

Sigma-Aldrich

Beta-galactoside alpha-2,6-sialyltransferase 1

≥300 units/mg protein, ST6GAL1 human recombinant, expressed in HEK 293 cells

Synonym(s):

Alpha 2,6-ST 1, B-cell antigen CD75, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1, ST6Gal I, Sialyltransferase 1

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.54

recombinant

expressed in HEK 293 cells

description

The specific activity of ST6Gal I is measured by its ability to transfer sialic acid from CMP-NANA to asialofetuin.

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

specific activity

≥300 units/mg protein

shipped in

ambient

storage temp.

−20°C

General description

Recombinant human Beta-galactoside alpha-2,6-sialyltransferase 1 (ST6Gal I) is expressed in human HEK 293 cells as a glycoprotein with a calculated molecular mass of 43.5 kDa (amino acids 27-406). The DTT-reduced protein migrates as a ~50 kDa polypeptide on SDS-PAGE due to glycosylation. This protein is manufactured in human cells, with no serum. The human cells expression system allows human-like glycosylation and folding, and often supports higher specific activity of the protein. The protein is produced with no artificial tags.

Biochem/physiol Actions

ST6Gal I catalyzes the transfer of CMP-N-acetylneuraminate (CMP-sialic acid, CMP-NANA) to the β-D-galactosyl-1,4-N-acetyl-D-glucosaminyl termini on glycoproteins. Sialic acids are distributed in a variety of glycolipids and glycoproteins.1 The sialic acid that is added to a galactose (Gal) can be bound either to the hydroxyl attached to carbon-3 of Gal to form an α-2,3 glycosidic linkage, or to the hydroxyl group attached to carbon-6 to form an α-2,6 glycosidic linkage.1 ST6Gal I generates a α-2,6 linkage of sialic acid on the non-reducing, terminal Galβ1 4GlcNAc residues of oligosaccharides and glycoconjugates.2

Terminal sialylation has been shown to decrease Fcγ receptor binding and increase anti-inflammatory activity,3 as well as antibody-dependent cellular cytotoxicity in different studies by reduced binding of sialylated antibody towards FcγRIIIa.4-5

This recombinant ST6Gal I product can be used to study the mode of action of the enzyme, as well as its potential inhibitors. It can also be used as a glycoengineering tool to modify glycoproteins in vitro.

CMP-N-acetylneuraminate (CMP-sialic acid, CMP-NANA) to the β-D-galactosyl-1,4-N-acetyl-D-glucosaminyl termini on glycoproteins.
Sialic acids are distributed in a variety of glycolipids and glycoproteins. The sialic acid that is added to a galactose (Gal) can be bound either to the hydroxyl attached to carbon-3 of Gal to form an α-2,3 glycosidic linkage, or to the hydroxyl group attached to carbon-6 to form an α-2,6 glycosidic linkage. ST6Gal I generates a α-2,6 linkage of sialic acid on the non-reducing, terminal Galβ1 4GlcNAc residues of oligosaccharides and glycoconjugates.

Terminal sialylation has been shown to decrease Fcγ receptor binding and increase anti-inflammatory activity, as well as antibody-dependent cellular cytotoxicity in different studies by reduced binding of sialylated antibody towards FcγRIIIa.

Unit Definition

One unit is defined as the amount of enzyme required to transfer 1.0 nanomole of sialic acid from CMP-NANA to asialofetuin per minute at pH 6.0, 37oC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael F Naso et al.
mAbs, 2(5), 519-527 (2010-08-19)
Covalently-linked glycans on proteins have many functional roles, some of which are still not completely understood. Antibodies have a very specific glycan modification in the Fc region that is required for mediating immune effector functions. These Fc glycans are typically
Bernard J Scallon et al.
Molecular immunology, 44(7), 1524-1534 (2006-10-19)
Although it is now clear that certain Fc glycan structures on immunoglobulin G (IgG) antibodies (Abs) can have a dramatic influence on binding to selected Fcgamma receptors (FcgammaR) and on Fc-mediated immune functions, the effects of all known Fc glycan
Yoshikatsu Kaneko et al.
Science (New York, N.Y.), 313(5787), 670-673 (2006-08-05)
Immunoglobulin G (IgG) mediates pro- and anti-inflammatory activities through the engagement of its Fc fragment (Fc) with distinct Fcg receptors (FcgRs). One class of Fc-FcgR interactions generates pro-inflammatory effects of immune complexes and cytotoxic antibodies. In contrast, therapeutic intravenous gamma
Makoto Ogata et al.
BMC biotechnology, 9, 54-54 (2009-06-09)
Sialic acid is a deoxy uronic acid with a skeleton of nine carbons which is mostly found on cell surface in animals. This sialic acid on cell surface performs various biological functions by acting as a receptor for microorganisms, viruses
J Weinstein et al.
The Journal of biological chemistry, 257(22), 13835-13844 (1982-11-25)
A Gal beta 1 to 4GlcNAc alpha 2 to 6 sialyltransferse and a Gal beta 1 to 3(4)GlcNAc alpha 2 to 3 sialyltransferase have been purified 23,000- and 860,000-fold to homogeneity from Triton CF-54 extracts of rat liver membranes. The

Articles

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service