Skip to Content
Merck
All Photos(5)

Key Documents

A0812

Sigma-Aldrich

N-Acetylneuraminic acid

≥95% anhydrous basis, synthetic

Synonym(s):

5-Acetamido-3,5-dideoxy-D-glycero-D-galactononulosonic acid, Lactaminic acid, NAN, NANA, Sialic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO9
CAS Number:
Molecular Weight:
309.27
Beilstein:
1716283
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

type

Type IV-S

Assay

≥95% anhydrous basis

form

powder

color

white to off-white

mp

184-186  °C

solubility

H2O: ~50 mg/mL

storage temp.

−20°C

SMILES string

O[C@@]1(O[C@@]([C@@H]([C@H](C1)O)NC(C)=O)([H])[C@@H]([C@@H](CO)O)O)C(O)=O

InChI

1S/C11H19NO9/c1-4(14)12-7-5(15)2-11(20,10(18)19)21-9(7)8(17)6(16)3-13/h5-9,13,15-17,20H,2-3H2,1H3,(H,12,14)(H,18,19)/t5-,6+,7+,8+,9+,11-/m0/s1

InChI key

SQVRNKJHWKZAKO-PFQGKNLYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Acetylneuraminic acid (NANA, Neu5Ac) is a major component of glycoconjugates such as glycolipids, glycoproteins and proteoglycans (sialogylcoproteins) where it confers selective binding characteristics to the glycosylated component. Neu5Ac is used to study its biochemistry, metabolism and uptake in vivo and in vitro. Neu5Ac is used in the development of nanocarriers.

Biochem/physiol Actions

Both sialic acid and neuraminic acid are loosely used to refer to conjugates of neuraminic acid. N-Acetylneuraminic acid is often found as the terminal sugar of cell surface glycoproteins. Cell surface glycoproteins have important roles in cell recognition and interaction as well as in cell adhesion. Membrane glycoproteins are also important in tumor growth and metastases.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kavita Agarwal et al.
PLoS biology, 18(8), e3000788-e3000788 (2020-08-26)
Women with bacterial vaginosis (BV), an imbalance of the vaginal microbiome, are more likely to be colonized by potential pathogens such as Fusobacterium nucleatum, a bacterium linked with intrauterine infection and preterm birth. However, the conditions and mechanisms supporting pathogen
Kannan Tharakaraman et al.
Cell, 153(7), 1475-1485 (2013-06-12)
Of the factors governing human-to-human transmission of the highly pathogenic avian-adapted H5N1 virus, the most critical is the acquisition of mutations on the viral hemagglutinin (HA) to "quantitatively switch" its binding from avian to human glycan receptors. Here, we describe
Emma R Job et al.
PloS one, 8(3), e59623-e59623 (2013-04-02)
Members of the pentraxin family, including PTX3 and serum amyloid P component (SAP), have been reported to play a role in innate host defence against a range of microbial pathogens, yet little is known regarding their antiviral activities. In this
Fiona E Fleming et al.
Journal of virology, 88(8), 4558-4571 (2014-02-07)
N-acetyl- and N-glycolylneuraminic acids (Sia) and α2β1 integrin are frequently used by rotaviruses as cellular receptors through recognition by virion spike protein VP4. The VP4 subunit VP8*, derived from Wa rotavirus, binds the internal N-acetylneuraminic acid on ganglioside GM1. Wa
Xiaoli Xiong et al.
Nature, 499(7459), 496-499 (2013-06-22)
Of the 132 people known to have been infected with H7N9 influenza viruses in China, 37 died, and many were severely ill. Infection seems to have involved contact with infected poultry. We have examined the receptor-binding properties of this H7N9

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service