Skip to Content
Merck
All Photos(7)

Key Documents

SAB4503754

Sigma-Aldrich

Anti-phospho-AMPK α (pThr172) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AMPK, Anti-AMPK2, Anti-AMPKa2, Anti-PRKAA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 62 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr172)

Gene Information

General description

AMP-activated protein kinase α (AMPKα) exists in two isoforms α1 and α2. The gene coding for AMPK α1 and AMPKα2 is mapped to human chromosome 5p11-p14 and 1p31 respectively. AMPKα is a 63kDa catalytic subunit of AMPK, involved in regulation of AMP levels. The AMP-activated protein kinase (AMPK) is a ubiquitously expressed cellular energy sensor.

Immunogen

The antiserum was produced against synthesized peptide derived from human AMPK alpha around the phosphorylation site of Thr172.

Immunogen Range: 140-189

Application

Anti-phospho-AMPK α (pThr172) antibody produced in rabbit has been used in western blot analysis.
Anti-phospho-AMPK α (pThr172) antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

AMP-activated protein kinase is activated by the increased concentration of adenosine mono phosphate (AMP) during mitochondrial dysfunction. AMPK, or AMPK α1, is a vital constituent of the adaptive response to cardiomyocyte stress that occurs during ischemia. AMPK is involved in regulation of various cellular processes such as mitochondrial biogenesis, autophagy and inflammation, and immune responses. AMPK signaling pathway aids in maintaining energy balance in retinal neurons, therefore, it is considered as a potential therapeutic target to prevent or delay retinal degeneration. AMPK acts as a cellular energy sensor that controls glucose and lipid metabolism in response to change in nutrient and intracellular energy levels. C allele of protein kinase AMP-activated catalytic subunit α 1 (PRKAA1) rs13361707 is associated with susceptibility of gastric cancer (GCa). AMPKα2 plays an essential role in controlling postmortem glycolysis in skeletal muscle.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activation of AMPK by metformin promotes renal cancer cell proliferation under glucose deprivation through its interaction with PKM2
Liu M, et al.
International Journal of Biological Sciences, 15(3), 617-617 (2019)
The AMPK signalling pathway coordinates cell growth, autophagy and metabolism.
Mihaylova MM and Shaw RJ
Nature Cell Biology, 13(9), 1016-1023 (2011)
AMPK in cardiac fibrosis and repair: Actions beyond metabolic regulation.
Daskalopoulos EP
Journal of Molecular and Cellular Cardiology, 91, 188-200 (2016)
Angiotensin-converting enzyme 2 regulates autophagy in acute lung injury through AMPK/mTOR signaling
Zhang X, et al.
Archives of Biochemistry and Biophysics, 672, 108061-108061 (2019)
Genetic variant of PRKAA1 and gastric cancer risk in an eastern Chinese population.
Qiu LX
Oncotarget, 6(40), 42661-42666 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service