Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200082

Sigma-Aldrich

Monoclonal Anti-Maltose Binding Protein (MBP) antibody produced in rat

1.0 mg/mL, clone MBP 7G4, purified immunoglobulin

Synonym(s):

Anti-MBP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MBP 7G4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~42 kDa

species reactivity

wide range

concentration

1.0 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 0.1-0.2 μg/mL using MBP recombinant protein

isotype

IgG2a

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

MBP is part of a large class of proteins that aid in the uptake of small molecules. While it naturally resides in the periplasm, MBP can also be expressed at high yields in the cytoplasm. For different proteins, increased solubility, enhanced stability and markedly improved yields have been reported after fusion to MBP.
Monoclonal Anti-Maltose Binding Protein (MBP) (rat IgG2a isotype) is derived from the hybridoma MBP 7G4 produced by the fusion of mouse myeloma cells (P3X63Ag8.653) and splenocytes from rat immunized with MBP-fusion protein.1 The antibody is purified from culture supernatant of hybridoma cells grown in a bioreactor. Monoclonal Anti- Maltose Binding Protein (MBP) is specific for MBP.

Specificity

Monoclonal Anti- Maltose Binding Protein (MBP) is specific for MBP.

Immunogen

MBP-fusion protein

Application

Monoclonal Anti-Maltose Binding Protein (MBP) antibody produced in rat is suitable for:
  • immunoblotting
  • enzyme linked immunosorbent assay (ELISA)
  • immunoprecipitation

Monoclonal antibodies recognizing specifically MBP are useful in various immunotechniques for identifying the expression of an MBP fusion protein in bacteria or in cells transfected with MBP fusion protein expressing vectors.

Biochem/physiol Actions

Maltose binding protein (MBP) tag is known to be used in genetic engineering to create a stable fusion product that does not appear to interfere with the bioactivity of the protein of interest or with the biodistribution of the MBP tagged product. The expression of polypeptides in-frame with maltose binding protein (MBP) allows for their easy purification from bacterial extracts under mild conditions, which employ a single affinity chromatographic step on amylose resin. This system and others based on the expression of fusion proteins utilize a specific protease cleaving site to facilitate correct cleavage of the fusion protein. Thus, the MBP system incorporates a factor Xa cleavage site at the carboxy terminus of the MBP sequence, and cleavage by factor Xa separates MBP from its partner protein. Many recombinant proteins have been engineered with MBP tags to facilitate the detection, isolation and purification of the proteins.

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at -20 °C. For continuous use, the product may be stored at 2-8 °C for up to one month. For extended use, freeze at -20 °Cin working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify by centrifugation. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative analyses of ubiquitin-like ATG8 and cysteine protease ATG4 autophagy genes in the plant lineage and cross-kingdom processing of ATG8 by ATG4
Seo E, et al.
Autophagy, 12(11), 2054-2068 (2016)
Overview of affinity tags for protein purification
Kimple ME, et al.
Current Protocols in Protein Science / Editorial Board, John E. Coligan ... [Et Al.], 73(1), 9-9 (2013)
José J Rodríguez-Herva et al.
Cellular microbiology, 14(5), 669-681 (2012-01-12)
The bacterial pathogen Pseudomonas syringae pv tomato DC3000 suppresses plant innate immunity with effector proteins injected by a type III secretion system (T3SS). The cysteine protease effector HopN1, which reduces the ability of DC3000 to elicit programmed cell death in
Fusion tags for protein solubility, purification and immunogenicity in Escherichia coli: the novel Fh8 system
Costa S, et al.
Frontiers in Microbiology, 5, 63-63 (2014)
A bacterial cysteine protease effector protein interferes with photosynthesis to suppress plant innate immune responses
Rodriguez HJ, et al.
Cellular Microbiology, 14(5), 669 ?681-669 ?681 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service