Skip to Content
Merck
All Photos(1)

Key Documents

CLS3603

Corning®  96 Well Black Polystyrene Microplate

flat bottom clear, black polystyrene plate (Ideal for fluorescent assays), Tissue Culture (TC)-treated surface, pkg of (individually wrapped), sterile, lid

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black polystyrene plate (Ideal for fluorescent assays)
flat bottom clear
polystyrene
round clear wells
size:
96 wells
surface area:
0.32 cm2 , cell growth area

cell growth area

cell growth area
well volume:
360 μL
well working volume:
75-200 μL

material

black polystyrene plate (Ideal for fluorescent assays)
flat bottom clear
polystyrene
round clear wells

sterility

sterile

feature

lid
skirt
plate format: 96 well standard

packaging

case of 48
pkg of (individually wrapped)

manufacturer/tradename

Corning 3603

size

96 wells

surface area

0.32 cm2 , cell growth area

cell growth area

cell growth area

well volume

360 μL

well working volume

75-200 μL

color

black

suitability

suitable for (fluorescent/luminescent assays)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Bottoms are 60% thinner than conventional polystyrene plates, resulting in lower background fluorescence and enabling readings down to 340 nm
  • Opaque walls to prevent well-to-well crosstalk
  • Optically clear flat well bottom permits direct microscopic viewing
  • Can be used for both top and bottom reading instruments
  • Flat bottoms with 360 μL total volume
  • Recommended working volumes of 75 to 200 μL
  • Nonreversable lids with condensation rings to reduce contamination
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Individually wrapped

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajesh Bhardwaj et al.
Science advances, 6(48) (2020-11-29)
Epithelial calcium channel TRPV6 plays vital roles in calcium homeostasis, and its dysregulation is implicated in multifactorial diseases, including cancers. Here, we study the molecular mechanism of selective nanomolar-affinity TRPV6 inhibition by (4-phenylcyclohexyl)piperazine derivatives (PCHPDs). We use x-ray crystallography and
Armin Holch et al.
Virulence, 11(1), 1402-1414 (2020-10-24)
The respiratory tract is a major entry site for microbial pathogens. To combat bacterial infections, the immune system has various defense mechanisms at its disposal, including antimicrobial peptides (AMPs). To search for novel AMPs from the respiratory tract, a peptide
Najoua Elhidar et al.
Journal of microbiological methods, 181, 106128-106128 (2020-12-22)
The NorA efflux pump is one of the most studied efflux systems in Staphylococcus aureus and confers multidrug resistance to a variety of dyes and antimicrobial compounds. Hence, inhibition of the NorA efflux pump might be a viable option for
Chungyong Han et al.
Molecular therapy oncolytics, 17, 293-305 (2020-05-06)
Antibody applications in cancer immunotherapy involve diverse strategies, some of which redirect T cell-mediated immunity via engineered antibodies. Affinity is a trait that is crucial for these strategies, as optimal affinity reduces unwanted side effects while retaining therapeutic function. Antibody-antigen pairs
Achille Schild et al.
International journal of molecular sciences, 21(16) (2020-08-09)
Calcium ions regulate a wide array of physiological functions including cell differentiation, proliferation, muscle contraction, neurotransmission, and fertilization. The endoplasmic reticulum (ER) is the major intracellular Ca2+ store and cellular events that induce ER store depletion (e.g., activation of inositol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service