Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500420

Sigma-Aldrich

Anti-RIPK1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-RIP, Anti-RIP1, Anti-Receptor-interacting serine/threonine-protein kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RIPK1(8737)

General description

Receptor interacting serine/threonine kinase 1 (RIPK1) gene is located at 6p25.2 on the human chromosome. It belongs to the RIPK family. RIPK1 is a necroptosis regulatory protein. It is made up of N-terminal kinase domain, a RIP homotypic interaction motif and its C-terminal contains the death domain.

Immunogen

RIPK1 antibody was raised against a 15 amino acid peptide from near the amino terminus of human RIPK1.

Application

Anti-RIPK1 antibody produced in rabbit has been used in:
  • western blotting
  • immunocytochemistry
  • immunofluorescence

Biochem/physiol Actions

Receptor interacting serine/threonine kinase 1 (RIPK1) is involved in inflammatory signaling and prosurvival. It is also involved in regulating the caspase-8-mediated apoptosis. RIPK1 plays a role in activating nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB) and mitogen-activated protein (MAP) kinases. Mutations in this gene is associated with impaired innate and adaptive immunity. It also leads to very early onset inflammatory bowel diseases (VEO-IBDs).
Receptor interacting serine/threonine kinase 1 (RIPK1) modulates cell survival and death. It also has a role in inflammation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500420.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nunzio Antonio Cacciola et al.
International journal of molecular sciences, 23(15) (2022-08-13)
Recent pharmacological research on milk whey, a byproduct of the dairy industry, has identified several therapeutic properties that could be exploited in modern medicine. In the present study, we investigated the anticancer effects of whey from Mediterranean buffalo (Bubalus bubalis)
RIPK1 regulates RIPK3-MLKL-driven systemic inflammation and emergency hematopoiesis
Rickard J A, et al.
Cell, 157(5), 1175-1188 (2014)
Tauroursodeoxycholic acid protects nucleus pulposus cells from compression-induced apoptosis and necroptosis via inhibiting endoplasmic reticulum stress
Wang W, et al.
Evidence-Based Complementary and Alternative Medicine : ECAM, 2018(3), 493-499 (2018)
Human RIPK1 deficiency causes combined immunodeficiency and inflammatory bowel diseases
Li Y, et al.
Proceedings of the National Academy of Sciences, 116(3), 970-975 (2019)
Xiang Li et al.
The Journal of surgical research, 225, 108-117 (2018-04-02)
Intestinal dysfunction, especially acute pathologies linked to intestinal ischemia/reperfusion (I/R) injury, is profoundly affected by inflammation and improper execution of cell death. Few studies have examined the efficacy of combined strategies in regulated intestinal epithelial necrosis after intestinal I/R. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service