Skip to Content
Merck
All Photos(3)

Key Documents

SAB2700986

Sigma-Aldrich

Anti-OPN3 (N-terminal) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

ECPN, OPN3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
western blot: 500-3000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... OPN3(23596)

Immunogen

Synthetic peptide corresponding to a region within amino acids 1 and 45 of Opsin 3 according to NP_055137

Application

Suggested starting dilutions are as follows: IHC-Fr: 1:100-1:1000, IHC-P: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Opsins are members of the guanine nucleotide-binding protein (G protein)-coupled receptor superfamily. In addition to the visual opsins, mammals possess several photoreceptive non-visual opsins that are expressed in extraocular tissues. This gene, opsin 3, is strongly expressed in brain and testis and weakly expressed in liver, placenta, heart, lung, skeletal muscle, kidney, and pancreas. The gene may also be expressed in the retina. The protein has the canonical features of a photoreceptive opsin protein. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiaki Yoshimoto et al.
Annals of gastroenterological surgery, 2(2), 154-161 (2018-06-05)
Light emitting-diodes (LED) have various effects on living organisms and recent studies have shown the efficacy of visible light irradiation from LED for anticancer therapies. However, the mechanism of LED's effects on cancer cells remains unclear. The aim of the
Bin Li et al.
Oncology letters, 11(3), 1693-1698 (2016-03-22)
While the anti-tumor actions of ginsenosides from Panax notoginseng are well-studied, the anti-proliferative activity of 20(S)-protopanaxadiol saponins (PDS) in Sanchi ginseng on human ovarian cancer has not been reported, nor has its effect on migration of SKOV3 cells been investigated.
Yuhei Waki et al.
Scientific reports, 13(1), 19954-19954 (2023-11-16)
Light-emitting diode (LED) irradiation has been used in the differentiation of mesenchymal stem cells into a variety of cell types. This study investigated the effect of green LED (GLED) irradiation on the differentiation of adipocyte-derived mesenchymal cells into hepatocyte-like cells
Toshiaki Yoshimoto et al.
BMC cancer, 24(1), 664-664 (2024-06-01)
Recent studies have shown that blue light-emitting diode (LED) light has anti-tumor effects, suggesting the possibility of using visible light in cancer therapy. However, the effects of blue light irradiation on cells in the tumor microenvironment, including tumor-associated macrophages (TAMs)
Toshiaki Yoshimoto et al.
Oncology reports, 47(5) (2022-04-14)
Irradiation with a specific wavelength of light using light‑emitting diodes (LEDs) has various effects on cells and organisms. Recently, the antitumor effects of visible blue light on tumor cells were reported; however, the mechanism and effects on the tumor microenvironment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service