Skip to Content
Merck
All Photos(1)

Key Documents

SIC001

Sigma-Aldrich

MISSION® siRNA Universal Negative Control #1

Synonym(s):

MISSION® siRNA Universal Negative Control

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51
Pricing and availability is not currently available.

product line

MISSION®

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

MISSION® siRNA Universal Negative Control #1 has been used as a negative control for siRNA transfections.[1][2][3]
Negative controls are an essential part of every siRNA experiment, and here we offer two sizes for your convenience. This negative control has been tested in vitro in Human, Mouse, and Rat cells.

Legal Information

All siRNA oligos are sold under license from the Massachusetts Institute of Technology.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoki Harada et al.
Journal of cellular biochemistry, 116(6), 998-1006 (2015-02-03)
Androgen receptor (AR) is known to bind to the same cis-element that glucocorticoid receptor (GR) binds to. However, the effects of androgen signaling on glucocorticoid signaling have not yet been elucidated. Here, we investigated the effects of testosterone on dexamethasone
Germline copy number variation of genes involved in chromatin remodelling in families suggestive of Li-Fraumeni syndrome with brain tumours.
Aury-Landas J et al.
European Journal of Human Genetics, 21, 1369-1369 (2013)
Adiponectin inhibits insulin function in primary trophoblasts by PPARa-mediated ceramide synthesis.
Aye IL et al.
Molecular Endocrinology, 28, 512-512 (2014)
Indrabahadur Singh et al.
BMC biology, 12, 21-21 (2014-03-26)
The high-mobility-group (HMG) proteins are the most abundant non-histone chromatin-associated proteins. HMG proteins are present at high levels in various undifferentiated tissues during embryonic development and their levels are strongly reduced in the corresponding adult tissues, where they have been
Shahanawaz Jiwani et al.
Infection and immunity, 80(3), 1252-1266 (2011-12-14)
The bacterial pathogen Listeria monocytogenes causes food-borne illnesses resulting in gastroenteritis, meningitis, or abortion. Listeria promotes its internalization into some human cells through binding of the bacterial surface protein InlB to the host receptor tyrosine kinase Met. The interaction of

Articles

MISSION siRNA Negative Controls ensure mRNA knockdown efficiency baseline; tested in human, rat, and mouse cells, designed without gene homology.

MISSION siRNA Negative Controls ensure mRNA knockdown efficiency baseline; tested in human, rat, and mouse cells, designed without gene homology.

MISSION siRNA Negative Controls ensure mRNA knockdown efficiency baseline; tested in human, rat, and mouse cells, designed without gene homology.

MISSION siRNA Negative Controls ensure mRNA knockdown efficiency baseline; tested in human, rat, and mouse cells, designed without gene homology.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service