Skip to Content
Merck
All Photos(1)

Key Documents

S9895

Sigma-Aldrich

Schneider′s Insect Medium

With L-glutamine, without calcium chloride and sodium bicarbonate, powder, suitable for insect cell culture

Synonym(s):

Schneider’s Drosophila Medium

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

Quality Level

form

powder

concentration

24.5 g/L

technique(s)

cell culture | insect: suitable

components

glucose: 2 g/L (Dextro)
NaHCO3: no
L-glutamine: 1.8 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Many insect tissue culture media were formulated to mimic the main physico-chemical properties of the body fluid of specific insects. A cursory survey of the formulas of culture media designed for insect tissues reveals great qualitative and quantitative disparities in composition. Different media designed for the same species may exhibit less similarity than do two media designed for insects of different orders. Various media have been devised for the in vitro culture of Drosophila cells and tissues. The most extensively used are Schneider′s medium and Echalier and Ohanessian′s D-22 medium. Drosophila cells have been employed to study a variety of biological processes including genetics, endocrinology, physiology and cell biology as well as recombinant protein expression.

Application

Originally developed for the culture of Drosophila; suitable for culture of other dipteran cell lines.
When supplemented with 5-20% heat-inactivated fetal bovine serum, Schneider′s medium has been found to support the rapid growth of both primary and established cultures of cells derived from Drosophila melanogaster. The medium has been used for the growth and maintenance of the cell lines originally derived by Schneider from Drosophila embryos as well as the culture of cells from other dipteran species.

SCHNEIDER′S INSECT MEDIUM is one of the cell culture media available from Sigma. The selection of a nutrient medium is strongly influenced by 1] type of cell, 2] type of culture [monolayer, suspension, clonal] and 3] ° of chemical definition necessary. It is important to review the literature for recommendations concerning medium, supplementation and physiological parameters required for a specific cell line.

Preparation Note

Powdered media are extremely hygroscopic and should be protected from atmospheric moisture. The entire contents of each package should be used immediately after opening. Preparing a concentrated solution of medium is not recommended as precipitates may form.

Supplements can be added prior to filtration or introduced aseptically to sterile medium. The nature of the supplement may affect storage conditions and shelf life of the medium.

1.Measure out 80% of final required volume of water. Water temperature should be 15-20 °C.
2.While gently stirring the water, add the powdered medium. Stir until dispersed. Material will not go in solution completely. Do NOT heat water.
3.Rinse original package with a small amount of water to remove all traces of powder. Add to solution in step 2.
4.To the solution in step 3, add 0.4 g sodium bicarbonate or 5.3 ml of sodium bicarbonate solution [7.5% w/v] for each liter of final volume of medium being prepared. Stir until dissolved.
5.While stirring, adjust the pH to at least 9.2 ± 0.2 with sodium hydroxide. Stir for a minimum of 10 minutes. Solution may become turbid.
6.While stirring, adjust the pH to 6.7 ± 0.2 with HCl, solution will clear.
7.Prepare a calcium chloride solution by dissolving 0.6 g of anhydrous calcium chloride [C5670] in 50 ml of tissue culture grade water for EACH liter of final volume of medium being prepared. Slowly add the calcium chloride solution dropwise to the medium with rapid mixing to avoid precipitate formation.
8.While stirring, adjust the pH of the medium to 0.1-0.3 pH units below the desired pH since it may rise during filtration. The use of 1N HCl or 1N NaOH is recommended.
9.Add additional water to bring the solution to final volume.
10.An osmotic pressure of 360 mOsm ± 5% is suitable for the growth of cells derived from Drosophila melanogaster. If desired the osmotic pressure can be increased 10 mOsm by adding potassium chloride (0.4 g salt OR 2 ml of a 20% (w/v) solution) OR sodium chloride (0.3 g of salt OR 2 ml of a 15% (w/v) solution) for EACH liter of final volume of medium being prepared. The osmotic pressure can be decreased 10 mOsm by adding 27.8 ml of water for EACH liter of final volume of medium being prepared. Stir until dissolved.
11.Sterilize immediately by filtration using a membrane with a porosity of 0.22 microns or less.
12.Aseptically dispense medium into sterile container.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alison Roth et al.
Scientific reports, 8(1), 12183-12183 (2018-08-17)
Malaria parasites transmitted by mosquito bite are remarkably efficient in establishing human infections. The infection process requires roughly 30 minutes and is highly complex as quiescent sporozoites injected with mosquito saliva must be rapidly activated in the skin, migrate through the
Xiao-Qing Ding et al.
Oncotarget, 8(47), 81953-81966 (2017-11-16)
Here, we found that ING5 overexpression resulted in a lower proliferation, reduced glucose metabolism, S arrest, decreased migration and invasion, apoptotic induction, fat accumulation, autophagy, senescence and mesenchymal-epithelial-transition of breast cancer cells. It also suppressed the tumor growth of breast
Xiao Zhang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(19), 4928-4941 (2017-04-21)
Phosphoinositides and their metabolizing enzymes are involved in Aβ
Aristeu Silva-Neto et al.
Revista da Sociedade Brasileira de Medicina Tropical, 53, e20200257-e20200257 (2020-11-12)
Biomphalaria snails may display varying levels of susceptibility to Schistosoma mansoni infection. We have been developing an in vitro model to study the interaction between the snail and the parasite, using tissue-derived cell cultures from Biomphalaria. The digestive gland- and
Weirong Jiang et al.
Journal of molecular cell biology, 11(9), 791-803 (2019-03-30)
Hedgehog (Hh) signalling plays conserved roles in controlling embryonic development; its dysregulation causes many diseases including cancers. The G protein-coupled receptor Smoothened (Smo) is the key signal transducer of the Hh pathway, whose posttranslational regulation has been shown to be

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service