Skip to Content
Merck
All Photos(1)

Key Documents

F4135

Sigma-Aldrich

Fetal Bovine Serum

USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma

Synonym(s):

HI FBS, HI FCS, HI sera, HI serum, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
eCl@ss:
32160409
NACRES:
NA.71
biological source:
bovine fetus
origin:
USA origin
sterility:
sterile-filtered
shipped in:
dry ice

biological source

bovine fetus

sterility

sterile-filtered

product line

FBS Classic

composition

Bovine IgG, ≤1 mg/mL
Hemoglobin, ≤20 mg/dL

origin

USA origin

technique(s)

cell culture | hybridoma: suitable
cell culture | insect: suitable
cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Fetal Bovine Serum (FBS) remains a popular media supplement as it provides a wide array of functions in cell culture. FBS delivers nutrients, growth and attachment factors and aids in proliferation. It also protects cells from oxidative damage and apoptosis. FBS blocks reactive oxygen species generation, which otherwise is not eliminated in serum-free media (SFM) systems.

Application

FBS sourced in the United States is used in a broad range of cell culture applications. FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture.
Fetal Bovine Serum has been used:
  • as a component of Dulbecco′s Modified Eagles Media (DME, DMEM), Eagle′s Minimum Essential Medium (EMEM), Roswell Park Memorial Institute medium (RPMI-1640), α-modified eagle medium (αMEM) (MEM) and Leibovitz L-15 medium
  • as a component of blocking medium for treating brainstem slices
  • to improve attachment of dermal explants

Preparation Note

Prepared by heating at 56°C for 30 minutes.
This product is prepared by heating at 56°C for 30 minutes. Heating serum to 56°C for 30 minutes inactivates the complement system for immunoassays. Heat activation also inactivates other undetermined inhibitors of cell growth in culture.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypoxia enhances secretion of matrix metalloproteinases in bovine dermal fibroblasts: an in vitro approach to bovine digital dermatitis
Akkoc A, et al.
Journal of Applied Animal Research, 46, 445-450 (2018)
Combining probiotic Phaeobacter inhibens DSM17395 and broad-host-range vibriophage KVP40 against fish pathogenic vibrios
Rasmussen BB, et al
Aquaculture (Amsterdam, Netherlands), 513, 734415-734415 (2019)
Antileishmanial activity study and theoretical calculations for 4-amino-1, 2, 4-triazole derivatives
Suleymanouglu N, et al.
Journal of Molecular Structure, 1144, 80-86 (2017)
Sanjay Ghosh et al.
Nucleic acids research, 44(9), e84-e84 (2016-02-07)
Long non-coding RNAs (lncRNAs) have emerged as regulators of gene expression across metazoa. Interestingly, some lncRNAs function independently of their transcripts - the transcription of the lncRNA locus itself affects target genes. However, current methods of loss-of-function analysis are insufficient
Ming Guan et al.
American journal of translational research, 11(3), 1343-1358 (2019-04-12)
Aging and exposure to noise or ototoxic drugs are major causes of hair cell death leading to human hearing loss, and many agents have been developed to protect hair cells from apoptosis. Fetal bovine serum (FBS) is a fundamental ingredient

Articles

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Calcium is an ionically stable divalent cation with important beneficial and toxic properties in cell culture. It is a component of a wide range of cell culture media.

Related Content

Learn how to effectively filter fetal bovine serum (FBS) using Stericup® Quick Release vacuum filtration devices, including flow test data and analysis.

Learn how to effectively filter fetal bovine serum (FBS) using Stericup® Quick Release vacuum filtration devices, including flow test data and analysis.

Learn how to effectively filter fetal bovine serum (FBS) using Stericup® Quick Release vacuum filtration devices, including flow test data and analysis.

Learn how to effectively filter fetal bovine serum (FBS) using Stericup® Quick Release vacuum filtration devices, including flow test data and analysis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service