Skip to Content
Merck
All Photos(1)

Key Documents

S9200

Sigma-Aldrich

Anti-SORL1 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-LDLR class A repeats containing, Anti-LR11, Anti-LRP9, Anti-SORLA, Anti-SorLA-1, Anti-Sortilin-related receptor, Anti-gp250

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~250 kDa

species reactivity

rat, human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using rat cerebellum (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SORL1(6653)
mouse ... Sorl1(20660)
rat ... Sorl1(300652)

General description

Sortilin related receptor 1 (SORL1), a multifunctional protein, is also known as SorLA-1, LR11, LRP9 and gp250. It is a type-I transmembrane protein highly expressed in neurons. This protein belongs to the low-density lipoprotein receptor family. It contains a transmembrane domain, a large extracellular domain and a short cytoplasmic tail. SORL1 gene is located on human chromosome 11q 23/24. This mosaic receptor contains a domain that shares similarity with the vacuolar protein sorting 10 (VPS10) and with the low-density lipoprotein receptor (LDLR) family of proteins.

Specificity

Anti-SORL1 (C-terminal) specifically recognizes rat SORL1.

Application

Anti-SORL1 (C-terminal) antibody produced in rabbit has been used in immunoblotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Sortilin related receptor 1 (SORL1) protein plays a key role in intracellular protein transport and endocytosis, cell signaling and the proliferation and migration of smooth muscle cells. It acts as a receptor for lipoproteins, glial cell-line-derived neurotrophic factor (GDNF) and platelet-derived growth factor (PDGF). Expression of this protein is developmentally regulated, with high transcript levels found during morphogenesis. SORLA is involved in development of atherosclerosis and in Alzheimer′s disease (AD). It mediates the uptake of apolipoprotein E (ApoE)-rich lipoproteins and is markedly induced in atheroma. SORL1 is known to play a regulating role in trafficking and processing of the amyloid precursor protein (APP) in neuronal cells. In the Golgi, SorLA/LR11 binding of APP leads to sequestration of APP and thereby it blocks the processing to the beta-amyloid peptide.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa Schmidt et al.
Cellular and molecular life sciences : CMLS, 74(8), 1475-1483 (2016-11-11)
Sorting-related receptor with A-type repeats (SORLA) is an intracellular sorting receptor that directs cargo proteins, such as kinases, phosphatases, and signaling receptors, to their correct location within the cell. The activity of SORLA assures proper function of cells and tissues
Satoru Suwa et al.
International heart journal, 61(3), 470-475 (2020-05-01)
Cardiovascular events still occur despite statin-based lipid-lowering therapy in patients with coronary artery disease (CAD). LR11, a member of the low-density lipoprotein receptor family, is a novel marker for the proliferation of intimal smooth muscle cells, which are critical to
Coding mutations in SORL 1 and A lzheimer disease
Vardarajan BN, et al.
Annals of Neurology, 77(2), 215-227 (2015)
L Jacobsen et al.
The Journal of biological chemistry, 271(49), 31379-31383 (1996-12-06)
The 39-40-kDa receptor-associated protein (RAP) binds to the members of the low density lipoprotein receptor gene family and functions as a specialized endoplasmic reticulum/Golgi chaperone. Using RAP affinity chromatography, we have purified a novel approximately 250-kDa brain protein and isolated
Olav M Andersen et al.
Biochemistry, 45(8), 2618-2628 (2006-02-24)
SorLA/LR11 is a sorting receptor that regulates the intracellular transport and processing of the amyloid precursor protein (APP) in neurons. SorLA/LR11-mediated binding results in sequestration of APP in the Golgi and in protection from processing into the amyloid-beta peptide (Abeta)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service