Skip to Content
Merck
All Photos(1)

Key Documents

C0413

Sigma-Aldrich

4-Chlorophenoxyacetic acid

BioReagent, suitable for plant cell culture, crystalline

Synonym(s):

4-CPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO3
CAS Number:
Molecular Weight:
186.59
Beilstein:
1211804
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Quality Level

form

crystalline

technique(s)

cell culture | plant: suitable

application(s)

agriculture

SMILES string

OC(=O)COc1ccc(Cl)cc1

InChI

1S/C8H7ClO3/c9-6-1-3-7(4-2-6)12-5-8(10)11/h1-4H,5H2,(H,10,11)

InChI key

SODPIMGUZLOIPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Chlorophenoxyacetic acid has been used in para-chlorophenoxyacetic acid (p-CPA) residue assay.
4-chlorophenoxy acetic acid (4-CPA), a chlorine derivative of phenoxyacetic acid (PA), is a plant growth regulator used as a herbicide.

Biochem/physiol Actions

4-Chlorophenoxyacetic acid is a synthetic auxin, that is used to induce parthenocarpic ovary growth on flowers of wild type plants.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomato fruit development in the auxin-resistant dgt mutant is induced by pollination but not by auxin treatment
Mignolli F, et al.
Journal of Plant Physiology, 169(12), 1165-1172 (2012)
p-CPA enhances growth and quality of muskmelon fruits
Hayata Y, et al.
Plant Growth Regulation, 36(1), 13-18 (2002)
G Carbonara et al.
Farmaco (Societa chimica italiana : 1989), 56(10), 749-754 (2001-11-23)
2-(4-Chloro-phenoxy)propanoic and 2-(4-chloro-phenoxy)butanoic acids are compounds known to block chloride membrane conductance in rat striated muscle by interaction with a specific receptor. In the present study, a series of chiral analogues has been prepared and tested to evaluate the influence
Ting-Fu Jiang et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 22(6), 811-814 (2006-06-15)
A new, simple and rapid capillary electrophoresis (CE) method, using hexadimethrine bromide (HDB) as electroosmotic flow (EOF) modifier, was developed for the identification and quantitative determination of four plant hormones, including gibberellin A3 (GA3), indole-3-acetic acid (IAA), alpha-naphthaleneacetic acid (NAA)
Ediz Yeşilkaya et al.
Hormone research, 72(4), 225-235 (2009-09-30)
Plant growth regulators are considered to leave minimal amounts of remnants and therefore cause no significant side effects in humans. In this study, we aimed to investigate the hormonal and histopathological effects of 4-chlorophenoxy acetic acid (4-CPA), a commonly used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service