Skip to Content
Merck
All Photos(2)

Key Documents

S6021

Sigma-Aldrich

D-Sorbitol

≥98% (GC), for molecular biology

Synonym(s):

D-Glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721899
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.31

grade

for molecular biology

vapor density

<1 (vs air)

vapor pressure

<0.1 mmHg ( 25 °C)

Assay

≥98% (GC)

form

powder or crystals

color

white

useful pH range

5.0-7 (25 °C, 182 g/L)

mp

98-100 °C (lit.)

solubility

water: soluble 182g/l at 20 °C (68 °F)

foreign activity

DNase, RNase, and protease, none detected

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-JGWLITMVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

May be used for washing spheroplasts and in isoelectric focusing to minimize endoosmotic flow in agarose gels. May be used to induce osmotic stress.

Biochem/physiol Actions

D-Sorbitol is a sugar alcohol that is commonly used as a sugar substitute. It occurs naturally and is also produced synthetically from glucose. The food industry uses D-sorbitol as an additive in the form of a sweetener, humectant, emulsifier, thickener, or dietary supplement. D-Sorbitol has also been found in cosmetics, paper, and pharmaceuticals. Naturally, D-sorbitol occurs widely in plants via photosynthesis, ranging from algae to higher order fruits of the family Rosaceae.

Other Notes

To gain a comprehensive understanding of our extensive range of Sugar alcohols for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isoelectric focusing.
D E Garfin
Methods in enzymology, 182, 459-477 (1990-01-01)
Jessica M Bryant et al.
Molecular systems biology, 16(8), e9569-e9569 (2020-08-21)
Mutually exclusive expression of the var multigene family is key to immune evasion and pathogenesis in Plasmodium falciparum, but few factors have been shown to play a direct role. We adapted a CRISPR-based proteomics approach to identify novel factors associated
Preparation of extracts from yeast.
S M Jazwinski
Methods in enzymology, 182, 154-174 (1990-01-01)
Kartik A Shah et al.
Biotechnology and bioengineering, 112(12), 2624-2629 (2015-06-03)
Monoclonal antibodies (mAbs) that bind and neutralize human pathogens have great therapeutic potential. Advances in automated screening and liquid handling have resulted in the ability to discover antigen-specific antibodies either directly from human blood or from various combinatorial libraries (phage
Arlene E Dent et al.
PloS one, 3(10), e3557-e3557 (2008-10-30)
Antibodies that impair Plasmodium falciparum merozoite invasion and intraerythrocytic development are one of several mechanisms that mediate naturally acquired immunity to malaria. Attempts to correlate anti-malaria antibodies with risk of infection and morbidity have yielded inconsistent results. Growth inhibition assays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service