Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
D-Sorbitol
Synonym(s):
D-Glucitol
Empirical Formula (Hill Notation):
C
6
H
14
O
6
CAS Number:
50-70-4
Molecular Weight:
182.17
EC Number:
200-061-5
Products
Product Category
carbohydrates (7)
sugar alcohols (4)
aroma chemicals (1)
Brand
Sigma-Aldrich (8)
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Optical Isomer
Special Grade
Food Grade (1)
Kosher (1)
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
S1876
D-Sorbitol,
≥98% (GC)
Expand
View Pricing
85529
D-Sorbitol,
BioUltra, ≥99.0% (HPLC)
Expand
View Pricing
240850
D-Sorbitol,
99% (GC)
Expand
View Pricing
S6021
D-Sorbitol,
≥98% (GC), for molecular biology
Expand
View Pricing
S3889
D-Sorbitol,
≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Expand
View Pricing
W302902
D-Sorbitol,
FCC, FG
Expand
View Pricing
S7547
D-Sorbitol,
≥98% (GC), BioXtra
Expand
View Pricing
97336
D-Sorbitol,
liquid, tested according to Ph. Eur.
Expand
View Pricing